製品名
TIMP-2 ヒト recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
製品番号
SRP3174
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
Appearancelyophilized powder
Colorwhite to off-white
Purity by SDS-Page≥95%
Purity by HPLC≥95%
Endotoxin Level≤1 EU/µg
Biological ActivityTIMP-2 activity was measured by its ability to inhibit human MMP-1 induced hydrolysis of a chromogenic peptide substrate at room temperature. Half maximal inhibition was obtained at a TIMP-2 concentration of approximately 0.5 µg/mL, when using an MMP-1 concentration of 1.6 µg/mL.