JP
JA
アプリケーション
製品
サービス
サポート
Home
Search Results
ラゾパニブ
(2)
ラゾパニブ
Empirical Formula (Hill Notation):
C
21
H
23
N
7
O
2
S
Molecular Weight:
437.52
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
CDS023580
ラゾパニブ
Expand
View Pricing
SML3104
ラゾパニブ, ≥98% (HPLC)
Expand
View Pricing