  • Search Results
  • The Protective Effect of Autophagy on DNA Damage in Mouse Spermatocyte-Derived Cells Exposed to 1800 MHz Radiofrequency Electromagnetic Fields.
The Protective Effect of Autophagy on DNA Damage in Mouse Spermatocyte-Derived Cells Exposed to 1800 MHz Radiofrequency Electromagnetic Fields.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-07-12)
Renyan Li, Mingfu Ma, Lianbing Li, Letian Zhao, Tianfeng Zhang, Xiaohan Gao, Danyan Zhang, Yijian Zhu, Qiang Peng, Xue Luo, Minglian Wang
ABSTRACT
The effects of exposure to radiofrequency electromagnetic fields (RF-EMFs) on the male reproductive system have raised public concern and studies have shown that exposure to RF-EMFs can induce DNA damage and autophagy. However, there are no related reports on the role of autophagy in DNA damage in spermatocytes, especially after exposure to RF-EMFs. The aim of the present study was to determine the mechanism and role of autophagy induced by RF-EMFs in spermatozoa cells. Mouse spermatocyte-derived cells (GC-2) were exposed to RF-EMFs 4 W/kg for 24 h. The level of reactive oxygen species (ROS) was determined by ROS assay kit. Comet assay was utilized to detect DNA damage. Autophagy was detected by three indicators: LC3II/LC3I, autophagic vacuoles, and GFP-LC3 dots, which were measured by western blot, transmission electron microscopy, and transfection with GFP-LC3, respectively. The expression of the molecular signaling pathway AMP-activated protein kinase (AMPK)/mTOR was determined by western blot. The results showed that RF-EMFs induced autophagy and DNA damage in GC-2 cells via ROS generation, and the autophagy signaling pathway AMPK/mTOR was activated by ROS generation. Furthermore, following inhibition of autophagy by knockdown of AMPKα, increased DNA damage was observed in GC-2 cells following RF-EMFs exposure, and overexpression of AMPKα promoted autophagy and attenuated DNA damage. These findings demonstrated that the autophagy which was induced by RF-EMFs via the AMPK/mTOR signaling pathway could prevent DNA damage in spermatozoa cells.
Sigma-Aldrich
抗LC3B ウサギ宿主抗体, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution