  • Search Results
  • Barbaloin protects against lipopolysaccharide (LPS)-induced acute lung injury by inhibiting the ROS-mediated PI3K/AKT/NF-κB pathway.
Barbaloin protects against lipopolysaccharide (LPS)-induced acute lung injury by inhibiting the ROS-mediated PI3K/AKT/NF-κB pathway.
International immunopharmacology (2018-09-03)
Kangfeng Jiang, Shuai Guo, Chao Yang, Jing Yang, Yu Chen, Aftab Shaukat, Gan Zhao, Haichong Wu, Ganzhen Deng
ABSTRACT
Barbaloin is the major anthraquinone compound that is isolated from the leaf exudates of Aloe vera and is often used as a bittering agent in alcoholic beverages. Here, we investigated the potential protective role of barbaloin in a mouse model of lipopolysaccharide (LPS)-induced acute lung injury (ALI) and clarified the underlying mechanism in vitro. Histological analysis showed that barbaloin exhibited a certain protective effect on LPS-induced ALI. To further elucidate the mechanisms underlying the actions of barbaloin, LPS-stimulated macrophages were used in this study. The results showed that barbaloin decreased the phosphorylation levels of IκBα and NF-κB p65, leading to a reduction in the expression of pro-inflammatory cytokines (TNF-α, IL-1β and IL-6). Furthermore, barbaloin also reduced the levels of intracellular reactive oxygen species (ROS) similarly to the antioxidant N‑acetyl‑l‑cysteine (NAC), which alone repressed the LPS-induced phosphorylation of phosphoinositide 3-kinase (PI3K) and AKT. Additionally, a pharmacological inhibitor of PI3K/AKT, LY294002, also restrained the phosphorylation levels of IκBα and NF-κB p65 and thereby decreased the expression of pro-inflammatory cytokines. Together, these results show that barbaloin possesses a protective effect on LPS-induced ALI via suppressing the release of pro-inflammatory cytokines through the ROS-mediated PI3K/AKT/NF-κB pathway.
Sigma-Aldrich
2′,7′-ジクロロフルオレセインジアセタート, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)