• ホーム
  • 検索結果
  • Resveratrol Decreases Oxidative Stress by Restoring Mitophagy and Improves the Pathophysiology of Dystrophin-Deficient mdx Mice.
Resveratrol Decreases Oxidative Stress by Restoring Mitophagy and Improves the Pathophysiology of Dystrophin-Deficient mdx Mice.
Oxidative medicine and cellular longevity (2018-12-05)
Rio Sebori, Atsushi Kuno, Ryusuke Hosoda, Takashi Hayashi, Yoshiyuki Horio
要旨
We previously showed that treatment with resveratrol (3,5,4'-trihydroxy-trans-stilbene), an activator of the NAD+-dependent deacetylase SIRT1 at 4 g/kg food for 32 weeks, significantly decreased the muscular reactive oxygen species (ROS) levels and ameliorated the pathology of mdx mice, an animal model of Duchenne muscular dystrophy (DMD). Here, we treated mdx mice with various doses of resveratrol (0.04, 0.4, and 4 g/kg food) for 56 weeks and examined the effects on serum creatine kinase levels and physical activities. Because resveratrol promotes autophagy, we also investigated whether autophagy including mitochondrial autophagy (mitophagy) is involved in resveratrol's effects. Autophagy/mitophagy-related genes and autophagic flux were downregulated in the muscle of mdx mice, and these phenomena were reversed by resveratrol with significant ROS reduction. Resveratrol at 4 g/kg food reduced the number of immature myofibers containing central nuclei and fine fibers < 400 μm2 and increased that of thicker myofibers in the quadriceps, suggesting that resveratrol decreased myofiber wasting and promoted muscular maturation. Accordingly, resveratrol at 0.4 g/kg food reduced the creatine kinase levels to one-third of those in untreated mdx mice and significantly increased the animals' physical activities. In C2C12 myoblast cells, resveratrol promoted mitophagy and eliminated mitochondria containing high superoxide levels. The clearance of damaged mitochondria and ROS reduction by resveratrol was completely suppressed by an autophagy inhibitor (chloroquine) and by knocking down Atg5 or Pink1, essential genes for autophagy and mitophagy, respectively. Thus, resveratrol is a potential therapeutic agent for DMD, and the clearance of damaged mitochondria probably contributes to its action.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
アンチマイシンA 放線菌由来
Sigma-Aldrich
クロロキン 二リン酸塩, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
モノクロナール抗α-チューブリン マウス宿主抗体, ascites fluid, clone B-5-1-2
Sigma-Aldrich
抗GAPDH抗体、マウスモノクローナル, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
セルリティック MT 細胞溶解試薬, For mammalian tissues