• ホーム
  • 検索結果
  • Effects of Treatment with the Hypomethylating Agent 5-aza-2'-deoxycytidine in Murine Type II Collagen-Induced Arthritis.
Effects of Treatment with the Hypomethylating Agent 5-aza-2'-deoxycytidine in Murine Type II Collagen-Induced Arthritis.
Pharmaceuticals (Basel, Switzerland) (2019-12-01)
Maria Cristina Petralia, Emanuela Mazzon, Maria Sofia Basile, Marco Cutuli, Roberto Di Marco, Fabiola Scandurra, Andrea Saraceno, Paolo Fagone, Ferdinando Nicoletti, Katia Mangano
要旨
The emerging role of epigenetics in the pathogenesis of autoimmune diseases has recently attracted much interest on the possible use of epigenetic modulators for the prevention and treatment of these diseases. In particular, we and others have shown that drugs that inhibit DNA methylation, such as azacitidine (AZA) and decitabine (DAC), already used for the treatment of acute myeloid leukemia, exert powerful beneficial effects in rodent models of type 1 diabetes, multiple sclerosis, and Guillain Barrè syndrome. Along this line of research, we have presently studied the effects of DAC in a murine model of rheumatoid arthritis induced by type II collagen and have demonstrated that DAC administration was associated with a significant amelioration of the clinical condition, along with in vivo and ex vivo modification of the immunological profile of the so-treated mice, that exhibited a diminished production of Th1 and Th17 pro-inflammatory cytokines and reduction of anti-type II collagen autoantibodies.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
α,α-Dicyanoethyl acetate, 98%