  • Search Results
  • AMPK-Independent LKB1 Activity Is Required for Efficient Epithelial Ovarian Cancer Metastasis.
AMPK-Independent LKB1 Activity Is Required for Efficient Epithelial Ovarian Cancer Metastasis.
Molecular cancer research : MCR (2019-11-21)
Adrian Buensuceso, Yudith Ramos-Valdes, Gabriel E DiMattia, Trevor G Shepherd
ABSTRACT
Epithelial ovarian cancer (EOC) spreads by direct dissemination of malignant cells and multicellular clusters, known as spheroids, into the peritoneum followed by implantation and growth on abdominal surfaces. Using a spheroid model system of EOC metastasis, we discovered that Liver kinase B1 (LKB1), encoded by the STK11 gene, and its canonical substrate AMP-activated protein kinase (AMPK) are activated in EOC spheroids, yet only LKB1 is required for cell survival. We have now generated STK11-knockout cell lines using normal human FT190 cells and three EOC cell lines, OVCAR8, HeyA8, and iOvCa147. STK11KO did not affect growth and viability in adherent culture, but it decreased anchorage-independent growth of EOC cells. EOC spheroids lacking LKB1 had markedly impaired growth and viability, whereas there was no difference in normal FT190 spheroids. To test whether LKB1 loss affects EOC metastasis, we performed intraperitoneal injections of OVCAR8-, HeyA8-, and iOvCa147-STK11KO cells, and respective controls. LKB1 loss exhibited a dramatic reduction on tumor burden and metastatic potential; in particular, OVCAR8-STK11KO tumors had evidence of extensive necrosis, apoptosis, and hypoxia. Interestingly, LKB1 loss did not affect AMPKα phosphorylation in EOC spheroids and tumor xenografts, indicating that LKB1 signaling to support EOC cell survival in spheroids and metastatic tumor growth occurs via other downstream mediators. We identified the dual-specificity phosphatase DUSP4 as a commonly upregulated protein due to LKB1 loss; indeed, DUSP4 knockdown in HeyA8-STK11KO cells partially restored spheroid formation and viability. IMPLICATIONS: LKB1 possesses key tumor-promoting activity independent of downstream AMPK signaling during EOC metastasis.
Sigma-Aldrich
オリゴマイシンA, ≥99% (HPLC)
Sigma-Aldrich
モノクロナール抗α-チューブリン マウス宿主抗体, clone B-5-1-2, ascites fluid
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
抗ビンキュリン抗体、モウスモノクローナル, clone hVIN-1, purified from hybridoma cell culture