  • Search Results
  • A novel chalcone derivative has antitumor activity in melanoma by inducing DNA damage through the upregulation of ROS products.
A novel chalcone derivative has antitumor activity in melanoma by inducing DNA damage through the upregulation of ROS products.
Cancer cell international (2020-02-06)
Keke Li, Shuang Zhao, Jing Long, Juan Su, Lisha Wu, Juan Tao, Jianda Zhou, JiangLin Zhang, Xiang Chen, Cong Peng
ABSTRACT
Melanoma is one of the most aggressive tumors with the remarkable characteristic of resistance to traditional chemotherapy and radiotherapy. Although targeted therapy and immunotherapy benefit advanced melanoma patient treatment, BRAFi (BRAF inhibitor) resistance and the lower response rates or severe side effects of immunotherapy have been observed, therefore, it is necessary to develop novel inhibitors for melanoma treatment. We detected the cell proliferation of lj-1-59 in different melanoma cells by CCK 8 and colony formation assay. To further explore the mechanisms of lj-1-59 in melanoma, we performed RNA sequencing to discover the pathway of differential gene enrichment. Western blot and Q-RT-PCR were confirmed to study the function of lj-1-59 in melanoma. We found that lj-1-59 inhibits melanoma cell proliferation in vitro and in vivo, induces cell cycle arrest at the G2/M phase and promotes apoptosis in melanoma cell lines. Furthermore, RNA-Seq was performed to study alterations in gene expression profiles after treatment with lj-1-59 in melanoma cells, revealing that this compound regulates various pathways, such as DNA replication, P53, apoptosis and the cell cycle. Additionally, we validated the effect of lj-1-59 on key gene expression alterations by Q-RT-PCR. Our findings showed that lj-1-59 significantly increases ROS (reactive oxygen species) products, leading to DNA toxicity in melanoma cell lines. Moreover, lj-1-59 increases ROS levels in BRAFi -resistant melanoma cells, leading to DNA damage, which caused G2/M phase arrest and apoptosis. Taken together, we found that lj-1-59 treatment inhibits melanoma cell growth by inducing apoptosis and DNA damage through increased ROS levels, suggesting that this compound is a potential therapeutic drug for melanoma treatment.
Sigma-Aldrich
(Tyr[SO3H]27)コレシストキニンフラグメント 26-33 アミド, ≥97% (HPLC), powder
Sigma-Aldrich
Anti-phospho-ATM (pSer1981) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)