  • Search Results
  • Pharmacological modulation of mitochondrial calcium uniporter controls lung inflammation in cystic fibrosis.
Pharmacological modulation of mitochondrial calcium uniporter controls lung inflammation in cystic fibrosis.
Science advances (2020-06-05)
Alessandro Rimessi, Chiara Pozzato, Lorenzo Carparelli, Alice Rossi, Serena Ranucci, Ida De Fino, Cristina Cigana, Anna Talarico, Mariusz R Wieckowski, Carla M P Ribeiro, Claudio Trapella, Giacomo Rossi, Giulio Cabrini, Alessandra Bragonzi, Paolo Pinton
ABSTRACT
Mitochondria physically associate with the endoplasmic reticulum to coordinate interorganelle calcium transfer and regulate fundamental cellular processes, including inflammation. Deregulated endoplasmic reticulum-mitochondria cross-talk can occur in cystic fibrosis, contributing to hyperinflammation and disease progression. We demonstrate that Pseudomonas aeruginosa infection increases endoplasmic reticulum-mitochondria associations in cystic fibrosis bronchial cells by stabilizing VAPB-PTPIP51 (vesicle-associated membrane protein-associated protein B-protein tyrosine phosphatase interacting protein 51) tethers, affecting autophagy. Impaired autophagy induced mitochondrial unfolding protein response and NLRP3 inflammasome activation, contributing to hyperinflammation. The mechanism by which VAPB-PTPIP51 tethers regulate autophagy in cystic fibrosis involves calcium transfer via mitochondrial calcium uniporter. Mitochondrial calcium uniporter inhibition rectified autophagy and alleviated the inflammatory response in vitro and in vivo, resulting in a valid therapeutic strategy for cystic fibrosis pulmonary disease.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
抗LC3B ウサギ宿主抗体, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
テトラメチルローダミンメチルエステルパークロレート, ≥95%
Sigma-Aldrich
モノクロナール抗アクチン マウス宿主抗体, clone AC-40, purified from hybridoma cell culture
Sigma-Aldrich
抗-MCU ウサギ宿主抗体, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
モノクローナル抗β-チューブリン マウス宿主抗体, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
KB-R7943, ≥98% (HPLC), powder