• ホーム
  • 検索結果
  • Triarylpyridine Compounds and Chloroquine Act in Concert to Trigger Lysosomal Membrane Permeabilization and Cell Death in Cancer Cells.
Triarylpyridine Compounds and Chloroquine Act in Concert to Trigger Lysosomal Membrane Permeabilization and Cell Death in Cancer Cells.
Cancers (2020-06-24)
Jennifer Beauvarlet, Rabindra Nath Das, Karla Alvarez-Valadez, Isabelle Martins, Alexandra Muller, Elodie Darbo, Elodie Richard, Pierre Soubeyran, Guido Kroemer, Jean Guillon, Jean-Louis Mergny, Mojgan Djavaheri-Mergny
要旨
Lysosomes play a key role in regulating cell death in response to cancer therapies, yet little is known on the possible role of lysosomes in the therapeutic efficacy of G-quadruplex DNA ligands (G4L) in cancer cells. Here, we investigate the relationship between the modulation of lysosomal membrane damage and the degree to which cancer cells respond to the cytotoxic effects of G-quadruplex ligands belonging to the triarylpyridine family. Our results reveal that the lead compound of this family, 20A promotes the enlargement of the lysosome compartment as well as the induction of lysosome-relevant mRNAs. Interestingly, the combination of 20A and chloroquine (an inhibitor of lysosomal functions) led to a significant induction of lysosomal membrane permeabilization coupled to massive cell death. Similar effects were observed when chloroquine was added to three new triarylpyridine derivatives. Our findings thus uncover the lysosomal effects of triarylpyridines compounds and delineate a rationale for combining these compounds with chloroquine to increase their anticancer effects.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
クロロキン 二リン酸塩, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Fluoromount水性マウンティング媒体, for use with fluorescent dye-stained tissues
Sigma-Aldrich
ヨウ化プロピジウム 溶液, solution (1.0 mg/ml in water)
Sigma-Aldrich
プロテアーゼインヒビターカクテル, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
フェニルメチルスルホニルフルオリド 溶液, ~0.1 M in ethanol (T)
Sigma-Aldrich
N-アセチル-L-システイン, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Q-VD-OPh 水和物, ≥95% (HPLC)
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
テトラメチルローダミンメチルエステルパークロレート, ≥95%
Sigma-Aldrich
ビスベンズイミドH 33258, for fluorescence, ≥98.0% (HPLC)
ブチルヒドロキシアニソール, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)