  • Search Results
  • Enzymatic properties and physiological function of glutamate racemase from Thermus thermophilus.
Enzymatic properties and physiological function of glutamate racemase from Thermus thermophilus.
Biochimica et biophysica acta. Proteins and proteomics (2020-06-01)
Tetsuya Miyamoto, Toshiyuki Moriya, Hiroshi Homma, Tairo Oshima
ABSTRACT
d-Amino acids are physiologically important components of peptidoglycan in the bacterial cell wall, maintaining cell structure and aiding adaptation to environmental changes through peptidoglycan remodelling. Therefore, the biosynthesis of d-amino acids is essential for bacteria to adapt to different environmental conditions. The peptidoglycan of the extremely thermophilic bacterium Thermus thermophilus contains d-alanine (d-Ala) and d-glutamate (d-Glu), but its d-amino acid metabolism remains poorly understood. Here, we investigated the enzyme activity and function of the product of the TTHA1643 gene, which is annotated to be a Glu racemase in the T. thermophilus HB8 genome. Among 21 amino acids tested, TTHA1643 showed highly specific activity toward Glu as the substrate. The catalytic efficiency (kcat/Km) of TTHA1643 toward d- and l-Glu was comparable; however, the kcat value was 18-fold higher for l-Glu than for d-Glu. Temperature and pH profiles showed that the racemase activity of TTHA1643 is high under physiological conditions for T. thermophilus growth. To assess physiological relevance, we constructed a TTHA1643-deficient strain (∆TTHA1643) by replacing the TTHA1643 gene with the thermostable hygromycin resistance gene. Growth of the ∆TTHA1643 strain in synthetic medium without d-Glu was clearly diminished relative to wild type, although the TTHA1643 deletion was not lethal, suggesting that alternative d-Glu biosynthetic pathways may exist. The deterioration in growth was restored by adding d-Glu to the culture medium, showing that d-Glu is required for normal growth of T. thermophilus. Collectively, our findings show that TTHA1643 is a Glu racemase and has the physiological function of d-Glu production in T. thermophilus.
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)