  • Search Results
  • Novel biphenyl diester derivative AB-38b inhibits NLRP3 inflammasome through Nrf2 activation in diabetic nephropathy.
Novel biphenyl diester derivative AB-38b inhibits NLRP3 inflammasome through Nrf2 activation in diabetic nephropathy.
Cell biology and toxicology (2019-11-27)
Lei Du, Jin Wang, Yibing Chen, Xizhi Li, Lei Wang, Yuan Li, Xiaoping Jin, Xiaoke Gu, Meng Hao, Xia Zhu, Xiaoxing Yin, Qian Lu
ABSTRACT
Inflammation reaction mediated by NLRP3 inflammasome and Nrf2-related oxidative stress are vital participants in the development of diabetic nephropathy (DN) and closely associated to kidney fibrosis. Nrf2, a known antioxidative transcription factor, has been reported to activate NLRP3 inflammasome through its downstream factors (HO-1, NQO1, etc.) recently. AB38b is a newly synthesized biphenyl diester derivative with a Nrf2 activation property. This research aims to evaluate the renal protective effects of AB-38b and to elucidate the anti-inflammation mechanisms involved. Type 2 diabetic mice induced by high fat diet with streptozocin (STZ) and high glucose-cultured mouse glomerular mesangial cells (GMCs) were used in current study. Results showed that administration of AB-38b improved the kidney function while attenuated renal fibrosis progression in diabetic mice together with reducing the extracellular matrix (ECM) accumulation of GMCs cultured in high glucose. Mechanistically, treatment with AB-38b significantly decreased the high level of NLRP3 inflammasome in diabetic condition by inhibiting the ROS/TXNIP/NLRP3 signaling pathway. And meanwhile, AB-38b treatment effectively improved Nrf2 signaling during diabetic condition. Furthermore, knocking down the gene expression of Nrf2 by siRNA in GMCs abolished the inhibition effect of AB-38b on NLRP3 inflammasome activation and ECM accumulation. Taken together, our data suggest that AB-38b was able to improve the renal function of diabetic mice, and the NLRP3 inflammasome inhibition effect of AB-38b was responsible for the renal protective effect. Further exploration indicate that Nrf2 plays pivotal role in AB-38b's attenuation of DN progression through inhibiting NLRP3 inflammasome activation.
Sigma-Aldrich
N-アセチル-L-システイン, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
ストレプトゾシン, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
D-(+)-グルコース, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
2',7'-ジクロロフルオレセイン二酢酸, ≥97%
Sigma-Aldrich
レスベラトロール, ≥99% (HPLC)
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)