• ホーム
  • 検索結果
  • VIP down-regulates the inflammatory potential and promotes survival of dying (neural crest-derived) corneal endothelial cells ex vivo: necrosis to apoptosis switch and up-regulation of Bcl-2 and N-cadherin.
VIP down-regulates the inflammatory potential and promotes survival of dying (neural crest-derived) corneal endothelial cells ex vivo: necrosis to apoptosis switch and up-regulation of Bcl-2 and N-cadherin.
Journal of neurochemistry (2009-03-03)
Shay-Whey M Koh, Jason Cheng, Rebecca M Dodson, Chao-Yar T Ku, Cara J Abbondandolo
要旨
The neuropeptide vasoactive intestinal peptide (VIP) is anti-inflammatory and protective in the immune and nervous systems, respectively. This study demonstrated in corneal endothelial (CE) cells injured by severe oxidative stress (1.4 mM H(2)O(2)) in bovine corneal organ cultures that VIP pre-treatment (0, 10(-10), 10(-8), and 10(-6) M; 15 min), in a VIP concentration-dependent manner, switched the inflammation-causing necrosis to inflammation-neutral apoptosis (showing annexin V-binding, chromatin condensation, and DNA fragmentation) and upheld ATP levels in a VIP antagonist (SN)VIPhyb-sensitive manner, while up-regulated mRNA levels of the anti-apoptotic Bcl-2 and the differentiation marker N-cadherin in a kinase A inhibitor-sensitive manner. As a result, VIP, in a concentration-dependent and VIP antagonist-sensitive manners, promoted long-term CE cell survival. ATP levels, a determining factor in the choice of apoptosis versus necrosis, measured after VIP pre-treatment and 0.5 min post-H(2)O(2) were 39.6 +/- 3.3, 50.8 +/- 6.2, 60.1 +/- 4.8, and 53.6 +/- 5.3 pmoles/microg protein (mean +/- SEM), respectively (p < 0.05, anova). VIP treatment alone concentration-dependently increased levels of N-cadherin (Koh et al. 2008), the phosphorylated cAMP-responsive-element binding protein and Bcl-2, while 10(-8) M VIP, in a VIP antagonist (SN)VIPhyb-sensitive manner, increased ATP level by 38% (p < 0.02) and decreased glycogen level by 32% (p < 0.02). VPAC1 (not VPAC2) receptor was expressed in CE cells. Thus, CE cell VIP/VPAC1 signaling is both anti-inflammatory and protective in the corneal endothelium.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, lyophilized, powder, ~70 U/mg
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, powder, white, ~120 U/mg
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, ≥260 U/mL, aqueous solution
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, Isoelectric focusing marker, pI 3.6
Sigma-Aldrich
アミログルコシダーゼ from Rhizopus sp., ≥40,000 units/g solid