  • Search Results
  • MicroRNA-195 controls MICU1 expression and tumor growth in ovarian cancer.
MicroRNA-195 controls MICU1 expression and tumor growth in ovarian cancer.
EMBO reports (2020-08-28)
Geeta Rao, Shailendra Kumar Dhar Dwivedi, Yushan Zhang, Anindya Dey, Khader Shameer, Ramachandran Karthik, Subramanya Srikantan, Md Nazir Hossen, Jonathan D Wren, Muniswamy Madesh, Joel T Dudley, Resham Bhattacharya, Priyabrata Mukherjee
ABSTRACT
MICU1 is a mitochondrial inner membrane protein that inhibits mitochondrial calcium entry; elevated MICU1 expression is characteristic of many cancers, including ovarian cancer. MICU1 induces both glycolysis and chemoresistance and is associated with poor clinical outcomes. However, there are currently no available interventions to normalize aberrant MICU1 expression. Here, we demonstrate that microRNA-195-5p (miR-195) directly targets the 3' UTR of the MICU1 mRNA and represses MICU1 expression. Additionally, miR-195 is under-expressed in ovarian cancer cell lines, and restoring miR-195 expression reestablishes native MICU1 levels and the associated phenotypes. Stable expression of miR-195 in a human xenograft model of ovarian cancer significantly reduces tumor growth, increases tumor doubling times, and enhances overall survival. In conclusion, miR-195 controls MICU1 levels in ovarian cancer and could be exploited to normalize aberrant MICU1 expression, thus reversing both glycolysis and chemoresistance and consequently improving patient outcomes.
Sigma-Aldrich
デオキシリボヌクレアーゼ I ウシ膵臓由来, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
ウシ血漿フィブロネクチン, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
ホルボール 12-ミリスタート 13-アセタート, ≥99% (TLC), film or powder
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)
Sigma-Aldrich
カルシウムイオノフォアA23187, ≥98% (TLC), powder
Sigma-Aldrich
乳酸アッセイキット, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
コラゲナーゼ Clostridium histolyticum由来, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture