• ホーム
  • 検索結果
  • Properties of retrograded and acetylated starch produced via starch extrusion or starch hydrolysis with pullulanase.
Properties of retrograded and acetylated starch produced via starch extrusion or starch hydrolysis with pullulanase.
Carbohydrate polymers (2013-08-06)
M Kapelko, T Zięba, A Gryszkin, M Styczyńska, A Wilczak
要旨
The aim of the present study was to determine the impact of serial modifications of starch, including firstly starch extrusion or hydrolysis with pullulanase, followed by retrogradation (through freezing and defrosting of pastes) and acetylation (under industrial conditions), on its susceptibility to amylolysis. The method of production had a significant effect on properties of the resultant preparations, whilst the direction and extent of changes depended on the type of modification applied. In the produced starch esters, the degree of substitution, expressed by the per cent of acetylation, ranged from 3.1 to 4.4 g/100 g. The acetylation had a significant impact on contents of elements determined with the atomic emission spectrometry, as it contributed to an increased Na content and decreased contents of Ca and K. The DSC thermal characteristics enabled concluding that the modifications caused an increase in temperatures and a decrease in heat of transition (or its lack). The acetylation of retrograded starch preparations increased their solubility in water and water absorbability. The modifications were found to exert various effects on the rheological properties of pastes determined based on the Brabender's pasting characteristics and flow curves determined with the use of an oscillatory-rotating viscosimeter. All starch acetates produced were characterized by ca. 40% resistance to amylolysis.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, lyophilized, powder, ~70 U/mg
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, powder, white, ~120 U/mg
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, ≥260 U/mL, aqueous solution
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, Isoelectric focusing marker, pI 3.6
Sigma-Aldrich
プルラナーゼ microbial
Sigma-Aldrich
アミログルコシダーゼ from Rhizopus sp., ≥40,000 units/g solid