• ホーム
  • 検索結果
  • Co-conjugation vis-à-vis individual conjugation of α-amylase and glucoamylase for hydrolysis of starch.
Co-conjugation vis-à-vis individual conjugation of α-amylase and glucoamylase for hydrolysis of starch.
Carbohydrate polymers (2013-08-31)
Swati B Jadhav, Rekha S Singhal
要旨
Two enzymes, α-amylase and glucoamylase have been individually and co-conjugated to pectin by covalent binding. Both the enzyme systems showed better thermal and pH stability over the free enzyme system with the complete retention of original activities. Mixture of individually conjugated enzymes showed lower inactivation rate constant with longer half life than the co-conjugated enzyme system. Individually conjugated enzymes showed an increase of 56.48 kJ/mole and 38.22 kJ/mole in activation energy for denaturation than the free enzymes and co-conjugated enzymes, respectively. Km as well as Vmax of individually and co-conjugated enzymes was found to be higher than the free enzymes. SDS-polyacrylamide gel electrophoresis confirmed the formation of conjugate and co-conjugate as evident by increased molecular weight. Both the enzyme systems were used for starch hydrolysis where individually conjugated enzymes showed highest release of glucose at 60 °C and pH 5.0 as compared to free and co-conjugated enzyme.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, lyophilized, powder, ~70 U/mg
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, powder, white, ~120 U/mg
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, ≥260 U/mL, aqueous solution
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, Isoelectric focusing marker, pI 3.6
Sigma-Aldrich
アミログルコシダーゼ from Rhizopus sp., ≥40,000 units/g solid