• ホーム
  • 検索結果
  • Carrier free co-immobilization of alpha amylase, glucoamylase and pullulanase as combined cross-linked enzyme aggregates (combi-CLEAs): a tri-enzyme biocatalyst with one pot starch hydrolytic activity.
Carrier free co-immobilization of alpha amylase, glucoamylase and pullulanase as combined cross-linked enzyme aggregates (combi-CLEAs): a tri-enzyme biocatalyst with one pot starch hydrolytic activity.
Bioresource technology (2013-09-04)
Sachin Talekar, Amol Pandharbale, Mayur Ladole, Shamraja Nadar, Mosin Mulla, Kshitija Japhalekar, Kishori Pattankude, Devika Arage
要旨
A tri-enzyme biocatalyst "combi-CLEAs" with starch hydrolytic activity was prepared from commercially available alpha amylase, glucoamylase and pullulanase preparations by aggregating enzymes with ammonium sulphate followed by cross-linking formed aggregates for 4.5h with 40 mM glutaraldehyde. The effects of precipitant type and cross-linking were studied and the biocatalyst was characterized. Scanning electron microscopy analysis showed that tri-enzyme biocatalyst was of spherical structure. For one pot starch hydrolytic activity, shift in optimum pH from 6 to 7 and temperature from 65 to 75 °C were observed after co-immobilization of enzymes. After one pot starch hydrolysis reaction in batch mode, 100%, 60% and 40% conversions were obtained with combi-CLEAs, separate CLEAs mixture and free enzyme mixture, respectively. Co-immobilization also enhanced the thermal stability of enzymes. Finally, the catalytic activity of enzymes in combi-CLEAs during one pot starch hydrolysis was well maintained up to five cycles without performance changes.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, lyophilized, powder, ~70 U/mg
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, powder, white, ~120 U/mg
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, ≥260 U/mL, aqueous solution
Sigma-Aldrich
アミログルコシダーゼ from Aspergillus niger, Isoelectric focusing marker, pI 3.6
Sigma-Aldrich
プルラナーゼ microbial
Sigma-Aldrich
アミログルコシダーゼ from Rhizopus sp., ≥40,000 units/g solid