• ホーム
  • 検索結果
  • The role of the 3' region of mammalian gonadotropin β subunit gene in the luteinizing hormone to chorionic gonadotropin evolution.
The role of the 3' region of mammalian gonadotropin β subunit gene in the luteinizing hormone to chorionic gonadotropin evolution.
Molecular and cellular endocrinology (2013-11-19)
Reut Gabay, Shelly Rozen, Albena Samokovlisky, Yehudit Amor, Rakefet Rosenfeld, Fortune Kohen, Abraham Amsterdam, Peter Berger, David Ben-Menahem
要旨
CGβ subunits comprise a unique carboxyl-terminal peptide (CTP) that has multiple O-linked glycans and extends serum half-life of the protein. It has evolved by incorporating a previously untranslated region of the LHβ gene into the reading frame. Although CTP-like sequences are encrypted in the LHβ genes of several mammals, the CGβ subunit developed only in primates and equids. To study this restriction in evolution, we examined whether the cryptic CTP decoded from the bovine LHβ gene (boCTP) possesses key characteristics of the human (h) CGβ-CTP. The boCTP does not impede several crucial aspects of hormone biosynthesis, but compared to the hCGβ-CTP, the stretch lacks O-glycans and determinants for circulatory survival. O-glycan deficiency and the associated incapacity to extend serum half-life is a major drawback of the boCTP. This may explain why LH did not evolve into CG in ruminants and consequently alternative mechanisms evolved to delay luteolysis early in gestation.
ソーシャルメディア
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
研究、開発、製造
私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates.All Rights Reserved.
当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。