  • Search Results
  • Vpu is the main determinant for tetraspanin downregulation in HIV-1-infected cells.
Vpu is the main determinant for tetraspanin downregulation in HIV-1-infected cells.
Journal of virology (2015-01-09)
Marie Lambelé, Herwig Koppensteiner, Menelaos Symeonides, Nathan H Roy, Jany Chan, Michael Schindler, Markus Thali
ABSTRACT
Tetraspanins constitute a family of cellular proteins that organize various membrane-based processes. Several members of this family, including CD81, are actively recruited by HIV-1 Gag to viral assembly and release sites. Despite their enrichment at viral exit sites, the overall levels of tetraspanins are decreased in HIV-1-infected cells. Here, we identify Vpu as the main viral determinant for tetraspanin downregulation. We also show that reduction of CD81 levels by Vpu is not a by-product of CD4 or BST-2/tetherin elimination from the surfaces of infected cells and likely occurs through an interaction between Vpu and CD81. Finally, we document that Vpu-mediated downregulation of CD81 from the surfaces of infected T cells can contribute to preserving the infectiousness of viral particles, thus revealing a novel Vpu function that promotes virus propagation by modulating the host cell environment. The HIV-1 accessory protein Vpu has previously been shown to downregulate various host cell factors, thus helping the virus to overcome restriction barriers, evade immune attack, and maintain the infectivity of viral particles. Our study identifies tetraspanins as an additional group of host factors whose expression at the surfaces of infected cells is lowered by Vpu. While the downregulation of these integral membrane proteins, including CD81 and CD82, likely affects more than one function of HIV-1-infected cells, we document that Vpu-mediated lowering of CD81 levels in viral particles can be critical to maintaining their infectiousness.
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Roche
X-tremeGENE HP DNA トランスフェクション試薬, High-performance polymer reagent for transfecting many cell lines
Sigma-Aldrich
エチレンジアミン四酢酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
エチレンジアミン四酢酸 二ナトリウム塩 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
エチレンジアミン四酢酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
フルオロセインイソチオシアナート, アイソマー I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
エチレンジアミン四酢酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
エチレンジアミン四酢酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
エチレンジアミン四酢酸, purified grade, ≥98.5%, powder
Supelco
塩化ナトリウム, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-グリセルアルデヒド 3-リン酸 溶液, 45-55 mg/mL in H2O
Sigma-Aldrich
フルオロセイン 5(6)-イソチオシアナート, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
エチレンジアミン四酢酸, 99.995% trace metals basis
Supelco
塩化ナトリウム, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
リン酸カルシウム, purum p.a., ≥96.0% (calc. as Ca3(PO4)2, KT)
Sigma-Aldrich
塩化ナトリウム, 99.999% trace metals basis
Sigma-Aldrich
塩化ナトリウム, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
エチレンジアミン四酢酸, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
塩化ナトリウム, SAJ first grade, ≥99.0%
Sigma-Aldrich
エチレンジアミン四酢酸, ≥98.0% (KT)