• ホーム
  • 検索結果
  • Bookmarking promoters in mitotic chromatin: poly(ADP-ribose)polymerase-1 as an epigenetic mark.
Bookmarking promoters in mitotic chromatin: poly(ADP-ribose)polymerase-1 as an epigenetic mark.
Nucleic acids research (2014-05-28)
Niraj Lodhi, Andrew V Kossenkov, Alexei V Tulin
要旨
Epigenetics are the heritable changes in gene expression or cellular phenotype caused by mechanisms other than changes in the underlying DNA sequence. After mitosis, it is thought that bookmarking transcription factors remain at promoters, regulating which genes become active and which remain silent. Herein, we demonstrate that poly(ADP-ribose)polymerase-1 (PARP-1) is a genome-wide epigenetic memory mark in mitotic chromatin, and we further show that the presence of PARP-1 is absolutely crucial for reactivation of transcription after mitosis. Based on these findings, a novel molecular model of epigenetic memory transmission through the cell cycle is proposed.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ジメチルスルホキシド, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, for molecular biology
Sigma-Aldrich
ジメチルスルホキシド, anhydrous, ≥99.9%
Sigma-Aldrich
ジメチルスルホキシド, ACS reagent, ≥99.9%
Sigma-Aldrich
ジメチルスルホキシド, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
ジメチルスルホキシド, suitable for HPLC, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, ReagentPlus®, ≥99.5%
Sigma-Aldrich
ノコダゾール, ≥99% (TLC), powder
Sigma-Aldrich
ジメチルスルホキシド, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
ジメチルスルホキシド, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
ジメチルスルホキシド, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
ジメチルスルホキシド, PCR Reagent
Sigma-Aldrich
ジメチルスルホキシド, puriss. p.a., dried, ≤0.02% water
Supelco
ジメチルスルホキシド, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
ジメチルスルホキシド, analytical standard
USP
ジメチルスルホキシド, United States Pharmacopeia (USP) Reference Standard
ジメチルスルホキシド, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Monoclonal Anti-PARP1 antibody produced in mouse, clone 3G4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ジメチルスルホキシド, JIS special grade, ≥99.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
ジメチルスルホキシド, suitable for HPLC
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5%
Sigma-Aldrich
ジメチルスルホキシド, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
モノクローナル抗PARP抗体 マウス宿主抗体, clone C-2-10, ascites fluid
Sigma-Aldrich
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-RLN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-PARP1, (N-terminal) antibody produced in mouse, clone 2C7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-H2AFX, (N-terminal) antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
ソーシャルメディア
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
研究、開発、製造
私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates.All Rights Reserved.
当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。