  • Search Results
  • Mechanisms of resistance to apoptosis in the human acute promyelocytic leukemia cell line NB4.
Mechanisms of resistance to apoptosis in the human acute promyelocytic leukemia cell line NB4.
Annals of hematology (2014-10-18)
I Gañán-Gómez, M C Estañ-Omaña, P Sancho, P Aller, M C Boyano-Adánez
ABSTRACT
Current frontline therapies have improved overall survival in acute promyelocytic leukemia (APL) patients to exceptional rates; however, relapse is still a problem among high-risk and old patients. Therefore, the development of better and safer therapies continues to be a goal in the treatment of this disease. In the present work, we examined three different pathways that hinder cell death in the APL cell line NB4, shedding light on the mechanisms that underlie resistance to apoptosis in these cells and that might help provide them with a proliferative advantage. We found that the proteasome inhibitor MG-132 specifically induces in NB4 cells an Nrf2-mediated antioxidant response which counteracts mitochondria-dependent apoptosis induced by the lipophilic cation dequalinium. More importantly, we also demonstrated that high basal autophagy levels and the gain-of-function of mutant p53 are intrinsic mechanisms of resistance to apoptosis in this cell line. According to our results, the pharmacological inhibition of autophagy and p53 mutants are useful tools to explore resistance to apoptosis in APL and other types of cancer and could be the bases of new therapeutic approaches that improve the efficiency and allow dose reduction of the current treatments.
Sigma-Aldrich
N-アセチル-L-システイン, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-アセチル-L-システイン, BioReagent, suitable for cell culture
Sigma-Aldrich
ヨウ化プロピジウム, ≥94.0% (HPLC)
Sigma-Aldrich
ヨウ化プロピジウム 溶液, solution (1.0 mg/ml in water)
Sigma-Aldrich
3-メチルアデニン, autophagy inhibitor
Supelco
ラパマイシン, VETRANAL®, analytical standard
Sigma-Aldrich
ラパマイシン, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
ローダミン123, mitochondrial specific fluorescent dye
Sigma-Aldrich
ヨウ化プロピジウム, ≥94% (HPLC)
Sigma-Aldrich
N-アセチル-L-システイン, BioXtra, ≥99% (TLC)
Supelco
N-アセチル-L-システイン, Pharmaceutical Secondary Standard; Certified Reference Material
USP
アセチルシステイン, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
ジヒドロエチジウム, ≥95%
Sigma-Aldrich
ジヒドロエチジウム, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
アクリジンオレンジ 塩酸塩 溶液, 10 mg/mL in H2O
Sigma-Aldrich
ローダミン123, BioReagent, for fluorescence, ≥85% (HPLC)
N-アセチル-L-システイン, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-アセチル-L-システイン, SAJ special grade, 98.0-102.0%
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)