  • Search Results
  • Carbamazepine attenuates inducible nitric oxide synthase expression through Akt inhibition in activated microglial cells.
Carbamazepine attenuates inducible nitric oxide synthase expression through Akt inhibition in activated microglial cells.
Pharmaceutical biology (2014-07-16)
Chen-Hsu Wang, Che-Jen Hsiao, Yi-Nung Lin, Joe-Wei Wu, Yu-Cheng Kuo, Ching-Kuo Lee, George Hsiao
ABSTRACT
Abstract Background: Carbamazepine, which was developed primarily for the treatment of epilepsy, is now also useful for the treatment of non-epileptic disorders and inflammatory hyperalgesia. However, the mechanism of its anti-neuroinflammatory action remains poorly understood. This study elucidates the anti-neuroinflammatory capacity of carbamazepine on microglial activation and the relative mechanisms involved. The microglial BV-2 cells were pretreated with carbamazepine for 15 min before activation by lipopolysaccharide (LPS). After LPS stimulation, the expression of inducible nitric oxide synthase (iNOS) was analyzed by Western blotting (WB) and reverse transcription-polymerase chain reaction. Signaling proteins and cyclooxygenase (COX)-2 were also evaluated by WB. The levels of nitrate and tumor necrosis factor (TNF)-α were analyzed by the Griess method and enzyme-linked immunosorbant assay, respectively. The formation of intracellular reactive oxygen species (ROS) was examined by fluorescent analysis. Carbamazepine strongly attenuated LPS-induced production of NO and iNOS protein at concentrations of 5, 10, and 20 μM. Consistently, it could markedly suppress iNOS mRNA expression stimulated by LPS. Among the signaling pathways, LPS-mediated IκBα degradation or JNK MAPK phosphorylation was not affected by carbamazepine. Interestingly, it was found that carbamazepine could concentration-dependently inhibit LPS-activated phospho-Akt expression. Nevertheless, LPS-induced ROS production was not affected by carbamazepine. Carbamazepine (20 μM) affected either COX-2 expression or TNF-α production induced by LPS with approximately 70% and 51% inhibition, respectively. Our findings showed that carbamazepine exerted selective inhibition on LPS-induced microglial iNOS expression through the down-regulation of Akt activation, and thus may play a pivotal role of anti-neuroinflammation in its therapeutic efficacy.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
ホルボール 12-ミリスタート 13-アセタート, ≥99% (TLC), film or powder
Sigma-Aldrich
ドデシル硫酸ナトリウム, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ACS reagent, ≥99.0%
Sigma-Aldrich
ホルボール 12-ミリスタート 13-アセタート, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
ピロ炭酸ジエチル, 96% (NT)
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
フェニルメチルスルホニルフルオリド, ≥98.5% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
フッ化ナトリウム, ACS reagent, ≥99%
Sigma-Aldrich
硝酸ナトリウム, ReagentPlus®, ≥99.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
HEPES緩衝液, 1 M in H2O
Sigma-Aldrich
硝酸ナトリウム, ACS reagent, ≥99.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 20% in H2O
SAFC
HEPES
Sigma-Aldrich
オルトバナジン酸ナトリウム, ≥90% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
ピロ炭酸ジエチル, 96% (NT)
Supelco
ドデシル硫酸ナトリウム, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
カルバマゼピン, powder
Sigma-Aldrich
フェニルメチルスルホニルフルオリド, ≥99.0% (T)
Supelco
ドデシル硫酸ナトリウム, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
オルトバナジン酸ナトリウム, 99.98% trace metals basis
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
ピロ炭酸ジエチル, 96% (NT)