  • Search Results
  • Unique roles of estrogen-dependent Pten control in epithelial cell homeostasis of mouse vagina.
Unique roles of estrogen-dependent Pten control in epithelial cell homeostasis of mouse vagina.
Oncogene (2014-03-19)
S Miyagawa, M Sato, T Sudo, G Yamada, T Iguchi
ABSTRACT
Numerous studies support a role of phosphatase and tensin homolog deleted from chromosome 10 (Pten) as a tumor suppressor gene that controls epithelial cell homeostasis to prevent tumor formation. Mouse vaginal epithelium cyclically exhibits cell proliferation and differentiation in response to estrogen and provides a unique model for analyzing homeostasis of stratified squamous epithelia. We analyzed vaginal epithelium-specific Pten conditional knockout (CKO) mice to provide new insights into Pten/phosphoinositide-3-kinase (PI3K)/Akt function. The vaginal epithelium of ovariectomized (OVX) mice (control) was composed of 1-2 layers of cuboidal cells, whereas OVX CKO mice exhibited epithelial hyperplasia in the suprabasal cells with increased cell mass and mucin production. This is possibly due to misactivation of mammalian target of rapamycin and mitogen-activated protein kinase. Intriguingly, estrogen administration to OVX Pten CKO mice induced stratification and keratinized differentiation in the vaginal epithelium, as in estrogen-treated controls. We found that Pten is exclusively expressed in the suprabasal cells in the absence of estrogens, whereas estrogen administration induced Pten expression in the basal cells. This suggests that Pten acts to prevent excessive cell proliferation as in the case of other squamous tissues. Thus, Pten exhibits a dual role on the control of vaginal homeostasis, depending on whether estrogens are present or absent. Our results provide new insights into how Pten functions in tissue homeostasis.
Sigma-Aldrich
ジメチルスルホキシド, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, anhydrous, ≥99.9%
Sigma-Aldrich
ジメチルスルホキシド, for molecular biology
Sigma-Aldrich
ジメチルスルホキシド, ACS reagent, ≥99.9%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
ジメチルスルホキシド, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
ジメチルスルホキシド, suitable for HPLC, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, ReagentPlus®, ≥99.5%
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
エチレンジアミン四酢酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
エチレンジアミン四酢酸 二ナトリウム塩 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, for molecular biology, ≥97.0%
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
ジメチルスルホキシド, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
ジメチルスルホキシド, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
エチレンジアミン四酢酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
HEPES緩衝液, 1 M in H2O
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, ≥97.0%
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
エチレンジアミン四酢酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
HEPES
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
ジメチルスルホキシド, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
エチレンジアミン四酢酸, anhydrous, crystalline, BioReagent, suitable for cell culture