Merck
TLR9 is critical for glioma stem cell maintenance and targeting.
Cancer research (2014-07-23)
Andreas Herrmann, Gregory Cherryholmes, Anne Schroeder, Jillian Phallen, Darya Alizadeh, Hong Xin, Tianyi Wang, Heehyoung Lee, Christoph Lahtz, Piotr Swiderski, Brian Armstrong, Claudia Kowolik, Gary L Gallia, Michael Lim, Christine Brown, Behnam Badie, Stephen Forman, Marcin Kortylewski, Richard Jove, Hua Yu
ABSTRACT
Understanding supports for cancer stem-like cells in malignant glioma may suggest therapeutic strategies for their elimination. Here, we show that the Toll-like receptor TLR9 is elevated in glioma stem-like cells (GSC) in which it contributes to glioma growth. TLR9 overexpression is regulated by STAT3, which is required for GSC maintenance. Stimulation of TLR9 with a CpG ligand (CpG ODN) promoted GSC growth, whereas silencing TLR9 expression abrogated GSC development. CpG-ODN treatment induced Frizzled4-dependent activation of JAK2, thereby activating STAT3. Targeted delivery of siRNA into GSC was achieved via TLR9 using CpG-siRNA conjugates. Through local or systemic treatment, administration of CpG-Stat3 siRNA to silence STAT3 in vivo reduced GSC along with glioma growth. Our findings identify TLR9 as a functional marker for GSC and a target for the delivery of efficacious therapeutics for glioma treatment. Cancer Res; 74(18); 5218-28. ©2014 AACR.
Sigma-Aldrich
メタノール, suitable for HPLC, ≥99.9%
Sigma-Aldrich
メタノール, anhydrous, 99.8%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
メタノール, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
メタノール, HPLC Plus, ≥99.9%
Sigma-Aldrich
メタノール, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
メタノール, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
HEPES緩衝液, 1 M in H2O
SAFC
HEPES
Supelco
メタノール, analytical standard
Sigma-Aldrich
メタノール, JIS special grade, ≥99.8%
Sigma-Aldrich
メタノール, Laboratory Reagent, ≥99.6%
Supelco
メタノール, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
D-ルシフェリン, synthetic
Sigma-Aldrich
メタノール, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
USP
メチルアルコール, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
メタノール, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
メタノール, BioReagent, ≥99.93%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SAFC
HEPES
Sigma-Aldrich
メタノール, Absolute - Acetone free
Sigma-Aldrich
メタノール, ACS reagent, ≥99.8%
Sigma-Aldrich
メタノール, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
メタノール, SAJ first grade, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
メタノール, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
メタノール, SAJ special grade
Sigma-Aldrich
メタノール, suitable for HPLC