  • Search Results
  • Mass Spectrometric Immunoassay for the qualitative and quantitative analysis of the cytokine Macrophage Migration Inhibitory Factor (MIF).
Mass Spectrometric Immunoassay for the qualitative and quantitative analysis of the cytokine Macrophage Migration Inhibitory Factor (MIF).
Proteome science (2014-10-21)
Nisha D Sherma, Chad R Borges, Olgica Trenchevska, Jason W Jarvis, Douglas S Rehder, Paul E Oran, Randall W Nelson, Dobrin Nedelkov
ABSTRACT
The cytokine MIF (Macrophage Migration Inhibitory Factor) has diverse physiological roles and is present at elevated concentrations in numerous disease states. However, its molecular heterogeneity has not been previously investigated in biological samples. Mass Spectrometric Immunoassay (MSIA) may help elucidate MIF post-translational modifications existing in vivo and provide additional clarity regarding its relationship to diverse pathologies. In this work, we have developed and validated a fully quantitative MSIA assay for MIF, and used it in the discovery and quantification of different proteoforms of MIF in serum samples, including cysteinylated and glycated MIF. The MSIA assay had a linear range of 1.56-50 ng/mL, and exhibited good precision, linearity, and recovery characteristics. The new assay was applied to a small cohort of human serum samples, and benchmarked against an MIF ELISA assay. The quantitative MIF MSIA assay provides a sensitive, precise and high throughput method to delineate and quantify MIF proteoforms in biological samples.
Sigma-Aldrich
アセトニトリル, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
アセトニトリル, anhydrous, 99.8%
Sigma-Aldrich
トルエン, anhydrous, 99.8%
Sigma-Aldrich
トリフルオロ酢酸, suitable for HPLC, ≥99.0%
Sigma-Aldrich
トリフルオロ酢酸, ReagentPlus®, 99%
Sigma-Aldrich
アジ化ナトリウム, ReagentPlus®, ≥99.5%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
1-メチル-2-ピロリジノン, anhydrous, 99.5%
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
D-(+)-グルコース, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
トルエン, ACS reagent, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
D-(+)-グルコース, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
デキストロース, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
1-メチル-2-ピロリジノン, ACS reagent, ≥99.0%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
トリフルオロ酢酸, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
アセトニトリル, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
アジ化ナトリウム, BioXtra
Sigma-Aldrich
D-(+)-グルコース, ACS reagent
Sigma-Aldrich
トルエン, suitable for HPLC, 99.9%
Sigma-Aldrich
アセトニトリル, ACS reagent, ≥99.5%
Sigma-Aldrich
トルエン, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
1-メチル-2-ピロリジノン, suitable for HPLC, ≥99%
Sigma-Aldrich
アセトニトリル, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
1-メチル-2-ピロリジノン, ReagentPlus®, 99%