• ホーム
  • 検索結果
  • FKBP5 expression in human adipose tissue increases following dexamethasone exposure and is associated with insulin resistance.
FKBP5 expression in human adipose tissue increases following dexamethasone exposure and is associated with insulin resistance.
Metabolism: clinical and experimental (2014-07-07)
Maria J Pereira, Jenny Palming, Maria K Svensson, Magnus Rizell, Jan Dalenbäck, Mårten Hammar, Tove Fall, Cherno O Sidibeh, Per-Arne Svensson, Jan W Eriksson
要旨
To study effects of dexamethasone on gene expression in human adipose tissue aiming to identify potential novel mechanisms for glucocorticoid-induced insulin resistance. Subcutaneous and omental adipose tissue, obtained from non-diabetic donors (10 M/15 F; age: 28-60 years; BMI: 20.7-30.6 kg/m²), was incubated with or without dexamethasone (0.003-3 μmol/L) for 24 h. Gene expression was assessed by microarray and real time-PCR and protein expression by immunoblotting. FKBP5 (FK506-binding protein 5) and CNR1 (cannabinoid receptor 1) were the most responsive genes to dexamethasone in both subcutaneous and omental adipose tissue (~7-fold). Dexamethasone increased FKBP5 gene and protein expression in a dose-dependent manner in both depots. The gene product, FKBP51 protein, was 10-fold higher in the omental than in the subcutaneous depot, whereas the mRNA levels were similar. Higher FKBP5 gene expression in omental adipose tissue was associated with reduced insulin effects on glucose uptake in both depots. Furthermore, FKBP5 gene expression in subcutaneous adipose tissue was positively correlated with serum insulin, HOMA-IR and subcutaneous adipocyte diameter and negatively with plasma HDL-cholesterol. FKBP5 SNPs were found to be associated with type 2 diabetes and diabetes-related phenotypes in large population-based samples. Dexamethasone exposure promotes expression of FKBP5 in adipose tissue, a gene that may be implicated in glucocorticoid-induced insulin resistance.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
モノクロナール抗β-アクチン マウス宿主抗体, clone AC-15, ascites fluid
Sigma-Aldrich
抗β-アクチン抗体, マウスモノクローナル, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
モノクロナール抗β-アクチン マウス宿主抗体, clone AC-74, ascites fluid
Sigma-Aldrich
アデノシン, ≥99%
Sigma-Aldrich
モノクロナール抗β-アクチン マウス宿主抗体, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
アデノシン, suitable for cell culture, BioReagent
Sigma-Aldrich
L-(−)-グルコース, ≥99%
Sigma-Aldrich
アデノシン
Supelco
アデノシン, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
抗β-アクチン ニワトリ宿主抗体, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
抗FKBP5抗体 ウサギ宿主抗体, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
アデノシン, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
抗FKBP4抗体 ウサギ宿主抗体, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
抗FKBP5抗体 マウス宿主抗体, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
抗FKBP4抗体 マウス宿主抗体, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-FKBP5 antibody produced in mouse, clone 3D10-1G11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
抗FKBP5抗体 ウサギ宿主抗体, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
抗FKBP5抗体 ウサギ宿主抗体, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
Sigma-Aldrich
抗FKBP5 ウサギ宿主抗体, affinity isolated antibody
Sigma-Aldrich
抗ACTB ウサギ宿主抗体, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-ACTB antibody produced in mouse, clone 3G4-F9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-FKBP4 antibody produced in mouse, clone 5C11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-FKBP5 antibody produced in mouse, clone 3D1-1B10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
抗FKBP4抗体 ウサギ宿主抗体, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
ソーシャルメディア
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
研究、開発、製造
私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates.All Rights Reserved.
当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。