  • Search Results
  • Muscle-specific Drp1 overexpression impairs skeletal muscle growth via translational attenuation.
Muscle-specific Drp1 overexpression impairs skeletal muscle growth via translational attenuation.
Cell death & disease (2015-02-27)
T Touvier, C De Palma, E Rigamonti, A Scagliola, E Incerti, L Mazelin, J-L Thomas, M D'Antonio, L Politi, L Schaeffer, E Clementi, S Brunelli
ABSTRACT
Mitochondrial fission and fusion are essential processes in the maintenance of the skeletal muscle function. The contribution of these processes to muscle development has not been properly investigated in vivo because of the early lethality of the models generated so far. To define the role of mitochondrial fission in muscle development and repair, we have generated a transgenic mouse line that overexpresses the fission-inducing protein Drp1 specifically in skeletal muscle. These mice displayed a drastic impairment in postnatal muscle growth, with reorganisation of the mitochondrial network and reduction of mtDNA quantity, without the deficiency of mitochondrial bioenergetics. Importantly we found that Drp1 overexpression activates the stress-induced PKR/eIF2α/Fgf21 pathway thus leading to an attenuated protein synthesis and downregulation of the growth hormone pathway. These results reveal for the first time how mitochondrial network dynamics influence muscle growth and shed light on aspects of muscle physiology relevant in human muscle pathologies.
Sigma-Aldrich
グリセロール, for molecular biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
グリセロール, ≥99.5%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ACS reagent, ≥99.0%
Sigma-Aldrich
グリセロール, ACS reagent, ≥99.5%
Sigma-Aldrich
グリセロール, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
DL-ジチオトレイトール 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
エチレンジアミン四酢酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
エチレンジアミン四酢酸 二ナトリウム塩 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Supelco
DL-ジチオトレイトール 溶液, 1 M in H2O
Sigma-Aldrich
カルボニルシアニド 4-(トリフルオロメトキシ)フェニルヒドラゾン, ≥98% (TLC), powder
Sigma-Aldrich
グリセロール, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
エチレンジアミン四酢酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
HEPES緩衝液, 1 M in H2O
Sigma-Aldrich
グリセリン, meets USP testing specifications
Sigma-Aldrich
グリセロール 溶液, 83.5-89.5% (T)
Supelco
グリセリン, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
エチレンジアミン四酢酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioXtra, ≥99.0% (GC)