• ホーム
  • 検索結果
  • Pharmacological identification of a guanidine-containing β-alanine analogue with low micromolar potency and selectivity for the betaine/GABA transporter 1 (BGT1).
Pharmacological identification of a guanidine-containing β-alanine analogue with low micromolar potency and selectivity for the betaine/GABA transporter 1 (BGT1).
Neurochemical research (2014-05-24)
Anas Al-Khawaja, Jette G Petersen, Maria Damgaard, Mette H Jensen, Stine B Vogensen, Maria E K Lie, Bolette Kragholm, Hans Bräuner-Osborne, Rasmus P Clausen, Bente Frølund, Petrine Wellendorph
要旨
The γ-aminobutyric acid (GABA) transporters (GATs) are key membrane transporter proteins involved in the termination of GABAergic signaling at synapses in the mammalian brain and proposed drug targets in neurological disorders such as epilepsy. To date, four different GAT subtypes have been identified: GAT1, GAT2, GAT3 and the betaine/GABA transporter 1 (BGT1). Owing to the lack of potent and subtype selective inhibitors of the non-GAT1 GABA transporters, the physiological role and therapeutic potential of these transporters remain to be fully understood. Based on bioisosteric replacement of the amino group in β-alanine or GABA, a series of compounds was generated, and their pharmacological activity assessed at human GAT subtypes. Using a cell-based [(3)H]GABA uptake assay, several selective inhibitors at human BGT1 were identified. The guanidine-containing compound 9 (2-amino-1,4,5,6-tetrahydropyrimidine-5-carboxylic acid hydrochloride) displayed more than 250 times greater potency than the parent compound β-alanine at BGT1 and is thus the most potent inhibitor reported to date for this subtype (IC50 value of 2.5 µM). In addition, compound 9 displayed about 400, 16 and 40 times lower inhibitory potency at GAT1, GAT2 and GAT3, respectively. Compound 9 was shown to be a substrate for BGT1 and to have an overall similar pharmacological profile at the mouse orthologue. Compound 9 constitutes an interesting pharmacological tool for specifically investigating the cellular pharmacology of BGT1 and is the first small-molecule substrate identified with such a high selectivity for BGT1 over the three other GAT subtypes.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
エタンスルホン酸, 95%
Sigma-Aldrich
グアニジン酢酸, 99%
Sigma-Aldrich
4-グアニジノ酪酸, ≥98%
Sigma-Aldrich
3-グアニジノプロピオン酸
Sigma-Aldrich
ニペコチン酸, 98%
Sigma-Aldrich
2-イミノ-1-イミダゾリジン酢酸, 98%
Sigma-Aldrich
エタンスルホン酸 溶液, 70 wt. % in H2O
ソーシャルメディア
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
研究、開発、製造
私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates.All Rights Reserved.
当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。