• ホーム
  • 検索結果
  • Role of LPAR3, PKC and EGFR in LPA-induced cell migration in oral squamous carcinoma cells.
Role of LPAR3, PKC and EGFR in LPA-induced cell migration in oral squamous carcinoma cells.
BMC cancer (2014-06-15)
Ingvild J Brusevold, Ingun H Tveteraas, Monica Aasrum, John Ødegård, Dagny L Sandnes, Thoralf Christoffersen
要旨
Oral squamous cell carcinoma is an aggressive neoplasm with serious morbidity and mortality, which typically spreads through local invasive growth. Lysophosphatidic acid (LPA) is involved in a number of biological processes, and may have a role in cancer cell migration and invasiveness. LPA is present in most tissues and can activate cells through six different LPA receptors (LPAR1-6). Although LPA is predominantly promigratory, some of the receptors may have antimigratory effects in certain cells. The signalling mechanisms of LPA are not fully understood, and in oral carcinoma cells the specific receptors and pathways involved in LPA-stimulated migration are unknown. The oral carcinoma cell lines E10, SCC-9, and D2 were investigated. Cell migration was studied in a scratch wound assay, and invasion was demonstrated in organotypic three dimensional co-cultures. Protein and mRNA expression of LPA receptors was studied with Western blotting and qRT-PCR. Activation of signalling proteins was examined with Western blotting and isoelectric focusing, and signalling mechanisms were further explored using pharmacological agents and siRNA directed at specific receptors and pathways. LPA stimulated cell migration in the two oral carcinoma cell lines E10 and SCC-9, but was slightly inhibitory in D2. The receptor expression profile and the effects of specific pharmacological antagonist and agonists indicated that LPA-stimulated cell migration was mediated through LPAR3 in E10 and SCC-9. Furthermore, in both these cell lines, the stimulation by LPA was dependent on PKC activity. However, while LPA induced transactivation of EGFR and the stimulated migration was blocked by EGFR inhibitors in E10 cells, LPA did not induce EGFR transactivation in SCC-9 cells. In D2 cells, LPA induced EGFR transactivation, but this was associated with slowing of a very high inherent migration rate in these cells. The results demonstrate LPA-stimulated migration in oral carcinoma cells through LPAR3, mediated further by PKC, which acts either in concert with or independently of EGFR transactivation.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ジメチルスルホキシド, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, for molecular biology
Sigma-Aldrich
ジメチルスルホキシド, anhydrous, ≥99.9%
Sigma-Aldrich
ジメチルスルホキシド, ACS reagent, ≥99.9%
Sigma-Aldrich
ジメチルスルホキシド, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
ジメチルスルホキシド, suitable for HPLC, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, ReagentPlus®, ≥99.5%
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
ヒドロコルチゾン, BioReagent, suitable for cell culture
Sigma-Aldrich
ジメチルスルホキシド, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
ヒドロコルチゾン, ≥98% (HPLC)
Sigma-Aldrich
ジメチルスルホキシド, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
ヒドロコルチゾン, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
抗GAPDH ウサギ宿主抗体, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
抗GAPDH抗体、マウスモノクローナル, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
ジメチルスルホキシド, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
プロスタグランジンE2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
オレオイル-L-α-リゾホスファチジン酸 ナトリウム塩, ≥98%, solid
Supelco
ヒドロコルチゾン, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
プロスタグランジンE2, ≥93% (HPLC), synthetic
Sigma-Aldrich
ジメチルスルホキシド, PCR Reagent
Sigma-Aldrich
CGS-12066 マレイン酸塩, solid
Sigma-Aldrich
ジメチルスルホキシド, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
プロスタグランジンE2, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
ジメチルスルホキシド, analytical standard
USP
ジメチルスルホキシド, United States Pharmacopeia (USP) Reference Standard
USP
ヒドロコルチゾン, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
ニューロテンシン, ≥90% (HPLC)
Sigma-Aldrich
ヒドロコルチゾン, meets USP testing specifications
ヒドロコルチゾン, European Pharmacopoeia (EP) Reference Standard