  • Search Results
  • Functional adaptation of presynaptic chemokine receptors in EAE mouse central nervous system.
Functional adaptation of presynaptic chemokine receptors in EAE mouse central nervous system.
Synapse (New York, N.Y.) (2014-08-06)
Silvia Di Prisco, Elisa Merega, Anna Pittaluga
ABSTRACT
In cortical synaptosomes of Experimental Autoimmune Encephalomyelitis (EAE) mice at the early stage of disease (13 days post immunization, d.p.i.), the Regulated upon Activation Normal T cell Expressed and Secreted (RANTES, CCL5)-mediated control of [3H]D-aspartate ([3H]D-ASP) exocytosis elicited by a mild depolarizing stimulus (12 mM KCl) shifted from inhibition to facilitation. By using selective antagonists for the chemokine receptor (CCR) 1, 3, and 5 subtypes, we found that the pharmacological profile of the receptor(s) accounting for CCL5-mediated effect was unaltered when compared to control. Inasmuch, CCR protein expression was unaltered. This studies was not extended at 21 d.p.i. since, at this stage, CCL5 failed to affect [3H]D-ASP exocytosis. At 13 d.p.i., the expression of CCR proteins was largely conserved when compared to control. In spinal cord synaptosomes of EAE mice at 21 d.p.i., when presynaptic defects became evident, the [3H]D-ASP exocytosis elicited by 15 mM KCl was significantly increased when compared to control and it was significantly potentiated by 1 nM CCL5. The antagonist pharmacological profile and the western blot analysis of the CCR proteins unveiled that the receptor repertoire involved was unmodified. Differently from controls, however, the CCR1 antagonist BX513 efficiently inhibited on its own [3H]D-ASP exocytosis suggesting that this receptor could have adopted an active conformation. Altogether, our observations favor the use of CCR antagonists to the cure of neurological symptoms in patients suffering from demyelinating syndrome.
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
塩化ナトリウム, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, for molecular biology, ~5 M in H2O
Supelco
塩化ナトリウム, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, 99.999% trace metals basis
Sigma-Aldrich
塩化ナトリウム, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
塩化ナトリウム, SAJ first grade, ≥99.0%
Sigma-Aldrich
塩化ナトリウム, tested according to Ph. Eur.
Sigma-Aldrich
塩化ナトリウム, tablet
Sigma-Aldrich
塩化ナトリウム, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
塩化ナトリウム 溶液, 1 M
Sigma-Aldrich
塩化ナトリウム, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
塩化ナトリウム, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
塩化ナトリウム 溶液, 0.1 M
Sigma-Aldrich
塩化ナトリウム-35Cl, 99 atom % 35Cl