  • Search Results
  • Bnip3 mediates doxorubicin-induced cardiac myocyte necrosis and mortality through changes in mitochondrial signaling.
Bnip3 mediates doxorubicin-induced cardiac myocyte necrosis and mortality through changes in mitochondrial signaling.
Proceedings of the National Academy of Sciences of the United States of America (2014-12-10)
Rimpy Dhingra, Victoria Margulets, Subir Roy Chowdhury, James Thliveris, Davinder Jassal, Paul Fernyhough, Gerald W Dorn, Lorrie A Kirshenbaum
ABSTRACT
Doxorubicin (DOX) is widely used for treating human cancers, but can induce heart failure through an undefined mechanism. Herein we describe a previously unidentified signaling pathway that couples DOX-induced mitochondrial respiratory chain defects and necrotic cell death to the BH3-only protein Bcl-2-like 19 kDa-interacting protein 3 (Bnip3). Cellular defects, including vacuolization and disrupted mitochondria, were observed in DOX-treated mice hearts. This coincided with mitochondrial localization of Bnip3, increased reactive oxygen species production, loss of mitochondrial membrane potential, mitochondrial permeability transition pore opening, and necrosis. Interestingly, a 3.1-fold decrease in maximal mitochondrial respiration was observed in cardiac mitochondria of mice treated with DOX. In vehicle-treated control cells undergoing normal respiration, the respiratory chain complex IV subunit 1 (COX1) was tightly bound to uncoupling protein 3 (UCP3), but this complex was disrupted in cells treated with DOX. Mitochondrial dysfunction induced by DOX was accompanied by contractile failure and necrotic cell death. Conversely, shRNA directed against Bnip3 or a mutant of Bnip3 defective for mitochondrial targeting abrogated DOX-induced loss of COX1-UCP3 complexes and respiratory chain defects. Finally, Bnip3(-/-) mice treated with DOX displayed relatively normal mitochondrial morphology, respiration, and mortality rates comparable to those of saline-treated WT mice, supporting the idea that Bnip3 underlies the cardiotoxic effects of DOX. These findings reveal a new signaling pathway in which DOX-induced mitochondrial respiratory chain defects and necrotic cell death are mutually dependent on and obligatorily linked to Bnip3 gene activation. Interventions that antagonize Bnip3 may prove beneficial in preventing mitochondrial injury and heart failure in cancer patients undergoing chemotherapy.
Sigma-Aldrich
ドキソルビシン 塩酸塩, 98.0-102.0% (HPLC)
Sigma-Aldrich
ドキソルビシン 塩酸塩, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
カルセイン AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
ドキソルビシン 塩酸塩, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
カルセイン AM 溶液, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
ジヒドロエチジウム, ≥95%
Sigma-Aldrich
テトラメチルローダミンメチルエステルパークロレート, ≥95%
Sigma-Aldrich
ジヒドロエチジウム, BioReagent, suitable for fluorescence, ≥95% (HPCE)
USP
ドキソルビシン 塩酸塩, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
カルセイン AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
Anti-UCP-3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution