  • Search Results
  • The Hmr and Lhr hybrid incompatibility genes suppress a broad range of heterochromatic repeats.
The Hmr and Lhr hybrid incompatibility genes suppress a broad range of heterochromatic repeats.
PLoS genetics (2014-03-22)
P R V Satyaki, Tawny N Cuykendall, Kevin H-C Wei, Nicholas J Brideau, Hojoong Kwak, S Aruna, Patrick M Ferree, Shuqing Ji, Daniel A Barbash
ABSTRACT
Hybrid incompatibilities (HIs) cause reproductive isolation between species and thus contribute to speciation. Several HI genes encode adaptively evolving proteins that localize to or interact with heterochromatin, suggesting that HIs may result from co-evolution with rapidly evolving heterochromatic DNA. Little is known, however, about the intraspecific function of these HI genes, the specific sequences they interact with, or the evolutionary forces that drive their divergence. The genes Hmr and Lhr genetically interact to cause hybrid lethality between Drosophila melanogaster and D. simulans, yet mutations in both genes are viable. Here, we report that Hmr and Lhr encode proteins that form a heterochromatic complex with Heterochromatin Protein 1 (HP1a). Using RNA-Seq analyses we discovered that Hmr and Lhr are required to repress transcripts from satellite DNAs and many families of transposable elements (TEs). By comparing Hmr and Lhr function between D. melanogaster and D. simulans we identify several satellite DNAs and TEs that are differentially regulated between the species. Hmr and Lhr mutations also cause massive overexpression of telomeric TEs and significant telomere lengthening. Hmr and Lhr therefore regulate three types of heterochromatic sequences that are responsible for the significant differences in genome size and structure between D. melanogaster and D. simulans and have high potential to cause genetic conflicts with host fitness. We further find that many TEs are overexpressed in hybrids but that those specifically mis-expressed in lethal hybrids do not closely correlate with Hmr function. Our results therefore argue that adaptive divergence of heterochromatin proteins in response to repetitive DNAs is an important underlying force driving the evolution of hybrid incompatibility genes, but that hybrid lethality likely results from novel epistatic genetic interactions that are distinct to the hybrid background.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
スクロース, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
塩化マグネシウム 溶液, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
スクロース, ≥99.5% (GC)
Sigma-Aldrich
スクロース, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
塩化マグネシウム, anhydrous, ≥98%
Sigma-Aldrich
塩化カルシウム 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
スクロース, BioUltra, for molecular biology, ≥99.5% (HPLC)
Supelco
スクロース, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
スクロース, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
HEPES緩衝液, 1 M in H2O
Sigma-Aldrich
塩化カルシウム, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
USP
スクロース, United States Pharmacopeia (USP) Reference Standard
SAFC
HEPES
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
スクロース, ACS reagent
Sigma-Aldrich
塩化マグネシウム 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
スクロース, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
スクロース, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
塩化カルシウム, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
塩化マグネシウム, powder, <200 μm
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
塩化マグネシウム 溶液, BioUltra, for molecular biology, 2 M in H2O
SAFC
HEPES
Sigma-Aldrich
塩化マグネシウム, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
塩化カルシウム, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
塩化マグネシウム 溶液, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)