• ホーム
  • 検索結果
  • Pigment epithelium-derived factor upregulates collagen I and downregulates matrix metalloproteinase 2 in osteosarcoma cells, and colocalises to collagen I and heat shock protein 47 in fetal and adult bone.
Pigment epithelium-derived factor upregulates collagen I and downregulates matrix metalloproteinase 2 in osteosarcoma cells, and colocalises to collagen I and heat shock protein 47 in fetal and adult bone.
The Journal of pharmacy and pharmacology (2014-09-02)
Marice B Alcantara, Natalie Nemazannikova, Mina Elahy, Crispin R Dass
要旨
Pigment epithelium-derived factor (PEDF) has proven anti-osteosarcoma activity. However, the mechanism(s) underpinning its ability to reduce primary bone tumour (osteosarcoma) metastasis is unknown. Adult and fetal murine bone were immunostained for PEDF, collagen I (major protein in bone) and its processing proteins, heat shock protein 47 (HSP47, a chaperone protein for collagen I), membrane type I matrix metalloproteinase (MT1-MMP, a collagenase), and matrix metalloproteinase 2 (MMP-2, which is activated by MT1-MMP). Immunoblotting and immunocytochemistry were used to observe levels of the above biomarkers when human osteosarcoma cells were treated with PEDF. Immunohistochemical staining in adult and fetal bone mirrors collagen I. PEDF localised to ridges of trabecular bone in tibial cortex and to megakaryocytes within bone marrow. Second, we observed that PEDF upregulates collagen I, HSP47 and MT1-MMP, while downregulating MMP-2 in osteosarcoma cells in vitro. PEDF is a promising antagonist to osteosarcoma cell metastasis via downregulation of MMP-2, and can induce tumour cells to further adopt differentiative properties, thereby possibly reducing their aggressive growth in vitro and in vivo.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
エタノール, SAJ first grade, ≥99.5%
Sigma-Aldrich
エチルアルコール(純粋), 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
エタノール, JIS special grade, ≥99.5%
Sigma-Aldrich
エタノール, ≥99.5%, SAJ super special grade
Sigma-Aldrich
エタノール, JIS first grade, 94.8-95.8%
Sigma-Aldrich
エタノール, ≥99.5%, suitable for HPLC
Sigma-Aldrich
エタノール, ≥99.5%
Sigma-Aldrich
エタノール, JIS special grade, 94.8-95.8%
Sigma-Aldrich
エタノール, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
エタノール, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
エタノール, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
エタノール, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
エタノール, puriss. p.a., ACS reagent, prima fine spirit, without additive, F15 o1, ~96%
Supelco
エタノール 溶液, certified reference material, 2000 μg/mL in methanol
Supelco
エタノール標準品10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
エタノール, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
エタノール, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
抗SERPINF1抗体 ウサギ宿主抗体, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
抗SERPINF1抗体 ウサギ宿主抗体, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
ソーシャルメディア
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
研究、開発、製造
私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates.All Rights Reserved.
当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。