SCF/C-KIT signaling modulates tryptase expression in acute myeloid leukemia cells.
International journal of hematology (2014-05-09)
Jingchun Jin, Yan Li, Yue Wang, Pingping Wang, Yazhu Wang
要旨
Tryptase is a serine protease with a variety of biological functions. Recently, elevated serum tryptase has been detected in certain patients with acute myeloid leukemia (AML). However, the underlying mechanism for the regulation of tryptase expression remains elusive. In this study, we aimed to investigate the role of stem cell factor (SCF)/C-KIT signaling in regulating the expression of tryptase in AML cells. We found a significant positive correlation between tryptase and C-KIT expression levels in AML patients. Furthermore, real-time PCR, Western blot and ELISA analysis showed that SCF upregulated tryptase mRNA and protein expression in U937 cells, and that this effect was abolished by pretreatment with PD98059 and SB230580. In addition, levels of phosphorylated ERK1/2 and p38MAPK correlated with tryptase levels. Taken together, these data suggest that the expression of tryptase is regulated by SCF/C-KIT signaling via the ERK1/2 and p38MAPK pathways.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
フルオロセインイソチオシアナート, アイソマー I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
フルオロセイン 5(6)-イソチオシアナート, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
フルオロセイン 5(6)-イソチオシアナート, ≥90% (HPLC)
Sigma-Aldrich
フルオロセインイソチオシアナート, アイソマー I, ≥97.5% (HPLC)
Sigma-Aldrich
フルオロセインイソチオシアナート, アイソマー I, ≥97.5% (HPLC)
Sigma-Aldrich
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-MAP Kinase 2 (ERK-2) antibody produced in mouse, clone 1B3B9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
Sigma-Aldrich
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
ソーシャルメディア
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
研究、開発、製造
私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates.All Rights Reserved.
当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。