  • Search Results
  • Bipyridine: the most widely used ligand. A review of molecules comprising at least two 2,2'-bipyridine units.
Bipyridine: the most widely used ligand. A review of molecules comprising at least two 2,2'-bipyridine units.
Chemical Reviews (2000)
Kaes C, et al.