• ホーム
  • 検索結果
  • Interaction of Salmonella enterica Serovar Typhimurium with Intestinal Organoids Derived from Human Induced Pluripotent Stem Cells.
Interaction of Salmonella enterica Serovar Typhimurium with Intestinal Organoids Derived from Human Induced Pluripotent Stem Cells.
Infection and immunity (2015-05-13)
Jessica L Forbester, David Goulding, Ludovic Vallier, Nicholas Hannan, Christine Hale, Derek Pickard, Subhankar Mukhopadhyay, Gordon Dougan
要旨
The intestinal mucosa forms the first line of defense against infections mediated by enteric pathogens such as salmonellae. Here we exploited intestinal "organoids" (iHOs) generated from human induced pluripotent stem cells (hIPSCs) to explore the interaction of Salmonella enterica serovar Typhimurium with iHOs. Imaging and RNA sequencing were used to analyze these interactions, and clear changes in transcriptional signatures were detected, including altered patterns of cytokine expression after the exposure of iHOs to bacteria. S. Typhimurium microinjected into the lumen of iHOs was able to invade the epithelial barrier, with many bacteria residing within Salmonella-containing vacuoles. An S. Typhimurium invA mutant defective in the Salmonella pathogenicity island 1 invasion apparatus was less capable of invading the iHO epithelium. Hence, we provide evidence that hIPSC-derived organoids are a promising model of the intestinal epithelium for assessing interactions with enteric pathogens.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
エタノール, SAJ first grade, ≥99.5%
Sigma-Aldrich
エチルアルコール(純粋), 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
アジ化ナトリウム, ReagentPlus®, ≥99.5%
Sigma-Aldrich
エタノール, JIS special grade, ≥99.5%
Sigma-Aldrich
エタノール, ≥99.5%, SAJ super special grade
Sigma-Aldrich
エタノール, JIS first grade, 94.8-95.8%
Sigma-Aldrich
エタノール, ≥99.5%, suitable for HPLC
Sigma-Aldrich
エタノール, ≥99.5%
Sigma-Aldrich
エタノール, JIS special grade, 94.8-95.8%
Sigma-Aldrich
エタノール, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
エタノール, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
エタノール, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
エタノール, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
レチノイン酸, ≥98% (HPLC), powder
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
塩化マグネシウム, anhydrous, ≥98%
Sigma-Aldrich
塩化マグネシウム 溶液, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩酸 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
塩酸, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
塩化カルシウム 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
L-グルタミン, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Supelco
塩酸 溶液, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
アジ化ナトリウム, BioXtra
Sigma-Aldrich
L-グルタミン, ReagentPlus®, ≥99% (HPLC)