  • Search Results
  • PARP-1 promotes autophagy via the AMPK/mTOR pathway in CNE-2 human nasopharyngeal carcinoma cells following ionizing radiation, while inhibition of autophagy contributes to the radiation sensitization of CNE-2 cells.
PARP-1 promotes autophagy via the AMPK/mTOR pathway in CNE-2 human nasopharyngeal carcinoma cells following ionizing radiation, while inhibition of autophagy contributes to the radiation sensitization of CNE-2 cells.
Molecular medicine reports (2015-04-16)
Ze-Tan Chen, Wei Zhao, Song Qu, Ling Li, Xiao-Di Lu, Fang Su, Zhong-Guo Liang, Si-Yan Guo, Xiao-Dong Zhu
ABSTRACT
It was previously reported that poly-(adenosine diphosphate-ribose) polymerase-1 (PARP-1) regulated ionizing radiation (IR)-induced autophagy in CNE-2 human nasopharyngeal carcinoma cells. The present study aimed to investigate whether PARP-1-mediated IR-induced autophagy occurred via activation of the liver kinase B1 (LKB1)/adenosine monophosphate-activated protein kinase (AMPK)/mammalian target of rapamycin (mTOR) signaling pathway in CNE-2 cells. In addition, the effect of PARP-1 and AMPK inhibition on the radiation sensitization of CNE-2 cells was investigated. CNE-2 cells were treated with 10 Gy IR in the presence or absence of the AMPK activator 5-amino-1-β-D-ribofuranosyl-1H-imid-azole-4-carboxamide (AICAR). In addition, IR-treated CNE-2 cells were transfected with lentivirus-delivered small-hairpin RNA or treated with the AMPK inhibitor Compound C. Western blot analysis was used to assess the protein expression of PARP-1, phosphorylated (p)-AMPK, microtubule-associated protein 1 light chain 3 (LC3)-II and p-P70S6K. Cell viability and clone formation assays were performed to determine the effect of PARP-1 silencing and AMPK inhibition on the radiation sensitization of CNE-2 cells. The results showed that IR promoted PARP-1, p-AMPK and LC3-II protein expression as well as decreased p-P70S6K expression compared with that of the untreated cells. In addition, AICAR increased the expression of p-AMPK and LC3-II as well as decreased p-P70S6K expression compared with that of the IR-only group; however, AICAR did not increase PARP-1 expression. Furthermore, PARP-1 gene silencing decreased the expression of PARP-1, p-AMPK and LC3-II as well as increased p-P70S6K expression. Compound C decreased p-AMPK and LC3-Ⅱ expression as well as increased p-P70S6K expression; however, Compound C did not increase PARP-1 expression. Western blot analysis detected limited expression of p-LKB1 in all treatment groups. Cell viability and clone formation assays revealed that PARP-1 or AMPK inhibition reduced the proliferation of CNE-2 cells following IR. In conclusion, the present study demonstrated that PARP-1 promoted autophagy via the AMPK/mTOR pathway; in addition, PARP-1 or AMPK inhibition contributed to the radiation sensitization of CNE-2 cells following IR. However, it remains to be elucidated whether PARP-1 is an upstream mediator of the LKB1 pathway in CNE‑2 cells following IR.
Sigma-Aldrich
ジメチルスルホキシド, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, anhydrous, ≥99.9%
Sigma-Aldrich
ジメチルスルホキシド, for molecular biology
Sigma-Aldrich
ジメチルスルホキシド, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
フェニルメチルスルホニルフルオリド, ≥98.5% (GC)
Sigma-Aldrich
ジメチルスルホキシド, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
抗LC3B ウサギ宿主抗体, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ジメチルスルホキシド, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
ジメチルスルホキシド, PCR Reagent
Sigma-Aldrich
フェニルメチルスルホニルフルオリド, ≥99.0% (T)
Sigma-Aldrich
ジメチルスルホキシド, JIS special grade, ≥99.0%
Sigma-Aldrich
ジメチルスルホキシド, suitable for HPLC
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
ジメチルスルホキシド, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
ジメチルスルホキシド, SAJ first grade, ≥99.0%
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5%