  • Search Results
  • Ca2+ influx-mediated dilation of the endoplasmic reticulum and c-FLIPL downregulation trigger CDDO-Me-induced apoptosis in breast cancer cells.
Ca2+ influx-mediated dilation of the endoplasmic reticulum and c-FLIPL downregulation trigger CDDO-Me-induced apoptosis in breast cancer cells.
Oncotarget (2015-06-09)
Soo Ah Jeong, In Young Kim, A Reum Lee, Mi Jin Yoon, Hyeseong Cho, Jong-Soo Lee, Kyeong Sook Choi
ABSTRACT
The synthetic triterpenoid 2-cyano-3, 12-dioxooleana-1, 9(11)-dien-C28-methyl ester (CDDO-Me) is considered a promising anti-tumorigenic compound. In this study, we show that treatment with CDDO-Me induces progressive endoplasmic reticulum (ER)-derived vacuolation in various breast cancer cells and ultimately kills these cells by inducing apoptosis. We found that CDDO-Me-induced increases in intracellular Ca2+ levels, reflecting influx from the extracellular milieu, make a critical contribution to ER-derived vacuolation and subsequent cell death. In parallel with increasing Ca2+ levels, CDDO-Me markedly increased the generation of reactive oxygen species (ROS). Interestingly, there exists a reciprocal positive-regulatory loop between Ca2+ influx and ROS generation that triggers ER stress and ER dilation in response to CDDO-Me. In addition, CDDO-Me rapidly reduced the protein levels of c-FLIPL (cellular FLICE-inhibitory protein) and overexpression of c-FLIPL blocked CDDO-Me-induced cell death, but not vacuolation. These results suggest that c-FLIPL downregulation is a key contributor to CDDO-Me-induced apoptotic cell death, independent of ER-derived vacuolation. Taken together, our results show that ER-derived vacuolation via Ca2+ influx and ROS generation as well as caspase activation via c-FLIPL downregulation are responsible for the potent anticancer effects of CDDO-Me on breast cancer cells.
Sigma-Aldrich
L-グルタチオン、還元型, ≥98.0%
Sigma-Aldrich
N-アセチル-L-システイン, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
チアゾリルブルーテトラゾリウムブロミド, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
チアゾリルブルーテトラゾリウムブロミド, 98%
Sigma-Aldrich
N-アセチル-L-システイン, BioReagent, suitable for cell culture
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, for molecular biology, ≥97.0%
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, ≥97.0%
Sigma-Aldrich
L-グルタチオン、還元型, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
2-アミノエチル ジフェニルボリナート, 97%
Sigma-Aldrich
L-グルタチオン、還元型, BioXtra, ≥98.0%
Sigma-Aldrich
N-アセチル-L-システイン, BioXtra, ≥99% (TLC)
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, BioXtra, ≥97 .0%
Sigma-Aldrich
1,2-ビス(2-アミノフェノキシ)エタン-N,N,N′,N′-四酢酸, 98%
Sigma-Aldrich
N-アセチル-L-システイン, SAJ special grade, 98.0-102.0%
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)