  • Search Results
  • Orexin excites rat inferior vestibular nuclear neurons via co-activation of OX1 and OX 2 receptors.
Orexin excites rat inferior vestibular nuclear neurons via co-activation of OX1 and OX 2 receptors.
Journal of neural transmission (Vienna, Austria : 1996) (2014-11-06)
Lei Yu, Xiao-Yang Zhang, Zhang-Peng Chen, Qian-Xing Zhuang, Jing-Ning Zhu, Jian-Jun Wang
ABSTRACT
Orexin deficiency results in cataplexy, a motor deficit characterized by sudden loss of muscle tone, strongly indicating an active role of central orexinergic system in motor control. However, effects of orexin on neurons in central motor structures are still largely unknown. Our previous studies have revealed that orexin excites neurons in the cerebellar nuclei and lateral vestibular nucleus, two important subcortical motor centers for control of muscle tone. Here, we report that both orexin-A and orexin-B depolarizes and increases the firing rate of neurons in the inferior vestibular nucleus (IVN), the largest nucleus in the vestibular nuclear complex and holding an important position in integration of information signals in the control of body posture. TTX does not block orexin-induced excitation on IVN neurons, suggesting a direct postsynaptic action of the neuropeptide. Furthermore, bath application of orexin induces an inward current on IVN neurons in a concentration-dependent manner. SB334867 and TCS-OX2-29, specific OX1 and OX2 receptor antagonists, blocked the excitatory effect of orexin, and [Ala(11), D-Leu(15)]-orexin B, a selective OX2 receptor agonist, mimics the orexin-induced inward current on IVN neurons. qPCR and immunofluorescence results show that both OX1 and OX2 receptor mRNAs and proteins are expressed and localized in the rat IVN. These results demonstrate that orexin excites the IVN neurons by co-activation of both OX1 and OX2 receptors, suggesting that via the direct modulation on the IVN, the central orexinergic system may actively participate in the central vestibular-mediated postural and motor control.
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
スクロース, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
塩化マグネシウム 溶液, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
スクロース, ≥99.5% (GC)
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
スクロース, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
塩化マグネシウム, anhydrous, ≥98%
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
スクロース, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
スクロース, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
スクロース, ACS reagent
Sigma-Aldrich
塩化マグネシウム 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
スクロース, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
硫酸マグネシウム 溶液, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
硫酸マグネシウム, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
塩化ナトリウム, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
塩化マグネシウム, powder, <200 μm
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
塩化マグネシウム 溶液, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
塩化ナトリウム, 99.999% trace metals basis
Sigma-Aldrich
硫酸マグネシウム 溶液, BioUltra, for molecular biology
Sigma-Aldrich
塩化マグネシウム, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
塩化マグネシウム 溶液, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
塩化ナトリウム, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture