  • Search Results
  • Results of the International Validation of the in vivo rodent alkaline comet assay for the detection of genotoxic carcinogens: Individual data for 1,2-dibromoethane, p-anisidine, and o-anthranilic acid in the 2nd step of the 4th phase Validation Study under the JaCVAM initiative.
Results of the International Validation of the in vivo rodent alkaline comet assay for the detection of genotoxic carcinogens: Individual data for 1,2-dibromoethane, p-anisidine, and o-anthranilic acid in the 2nd step of the 4th phase Validation Study under the JaCVAM initiative.
Mutation research. Genetic toxicology and environmental mutagenesis (2015-07-28)
Hironao Takasawa, Rie Takashima, Kazunori Narumi, Kazufumi Kawasako, Akiko Hattori, Masayoshi Kawabata, Shuichi Hamada
ABSTRACT
As part of the Japanese Center for the Validation of Alternative Methods (JaCVAM)-initiative International Validation Study of an in vivo rat alkaline comet assay, we examined 1,2-dibromoethane (DBE), p-anisidine (ASD), and o-anthranilic acid (ANT) to investigate the effectiveness of the comet assay in detecting genotoxic carcinogens. Each of the three test chemicals was administered to 5 male Sprague-Dawley rats per group by oral gavage at 48, 24, and 3h before specimen preparation. Single cells were collected from the liver and glandular stomach at 3h after the final dosing, and the specimens prepared from these two organs were subjected to electrophoresis under alkaline conditions (pH>13). The percentage of DNA intensity in the comet tail was then assessed using an image analysis system. A micronucleus (MN) assay was also conducted using these three test chemicals with the bone marrow (BM) cells collected from the same animals simultaneously used in the comet assay, i.e., combination study of the comet assay and BM MN assay. A genotoxic (Ames positive) rodent carcinogen, DBE gave a positive result in the comet assay in the present study, while a genotoxic (Ames positive) non-carcinogen, ASD and a non-genotoxic (Ames negative) non-carcinogen, ANT showed negative results in the comet assay. All three chemicals produced negative results in the BM MN assay. While the comet assay findings in the present study were consistent with those obtained from the rodent carcinogenicity studies for the three test chemicals, we consider the positive result in the comet assay for DBE to be particularly meaningful, given that this chemical produced a negative result in the BM MN assay. Therefore, the combination study of the comet assay and BM MN assay is a useful method to detect genotoxic carcinogens that are undetectable with the BM MN assay alone.
Sigma-Aldrich
ジメチルスルホキシド, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, anhydrous, ≥99.9%
Sigma-Aldrich
ジメチルスルホキシド, for molecular biology
Sigma-Aldrich
ジメチルスルホキシド, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
ホルムアルデヒド 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
水酸化ナトリウム 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
ホルムアルデヒド 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
水酸化ナトリウム 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
エタノール, JIS special grade, ≥99.5%
Sigma-Aldrich
ジメチルスルホキシド, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
エチルアルコール(純粋), 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
エタノール, SAJ first grade, ≥99.5%
Sigma-Aldrich
ホルムアルデヒド 溶液, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
ジメチルスルホキシド, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
水酸化ナトリウム 溶液, 0.1 M
Sigma-Aldrich
塩化ナトリウム, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
ジメチルスルホキシド, PCR Reagent
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
水酸化ナトリウム 溶液, 1 M
Sigma-Aldrich
ホルムアルデヒド 溶液, meets analytical specification of USP, ≥34.5 wt. %