  • Search Results
  • Mitochondrial fusion provides an 'initial metabolic complementation' controlled by mtDNA.
Mitochondrial fusion provides an 'initial metabolic complementation' controlled by mtDNA.
Cellular and molecular life sciences : CMLS (2015-02-25)
Liang Yang, Qi Long, Jinglei Liu, Haite Tang, Yuxing Li, Feixiang Bao, Dajiang Qin, Duanqing Pei, Xingguo Liu
ABSTRACT
Heteroplasmic cells, harboring both mutant and normal mitochondrial DNAs (mtDNAs), must accumulate mutations to a threshold level before respiratory activity is affected. This phenomenon has led to the hypothesis of mtDNA complementation by inter-mitochondrial content mixing. The precise mechanisms of heteroplasmic complementation are unknown, but it depends both on the mtDNA nucleoid dynamics among mitochondria as well as the mitochondrial dynamics as influenced by mtDNA. We tracked nucleoids among the mitochondria in real time to show that they are shared after complete fusion but not 'kiss-and-run'. Employing a cell hybrid model, we further show that mtDNA-less mitochondria, which have little ATP production and extensive Opa1 proteolytic cleavage, exhibit weak fusion activity among themselves, yet remain competent in fusing with healthy mitochondria in a mitofusin- and OPA1-dependent manner, resulting in restoration of metabolic function. Depletion of mtDNA by overexpression of the matrix-targeted nuclease UL12.5 resulted in heterogeneous mitochondrial membrane potential (ΔΨm) at the organelle level in mitofusin-null cells but not in wild type. In this system, overexpression of mitofusins or application of the fusion-promoting drug M1 could partially rescue the metabolic damage caused by UL12.5. Interestingly, mtDNA transcription/translation is not required for normal mitochondria to restore metabolic function to mtDNA-less mitochondria by fusion. Thus, interplay between mtDNA and fusion capacity governs a novel 'initial metabolic complementation'.
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ACS reagent, ≥99.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 20% in H2O
Supelco
ドデシル硫酸ナトリウム, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥90%
Sigma-Aldrich
テトラメチルローダミンメチルエステルパークロレート, ≥95%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥98.0% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
ドデシル硫酸ナトリウム, SAJ special grade, ≥97.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, SAJ first grade, ≥95.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0%