• ホーム
  • 検索結果
  • Upregulation of miR-362-3p Modulates Proliferation and Anchorage-Independent Growth by Directly Targeting Tob2 in Hepatocellular Carcinoma.
Upregulation of miR-362-3p Modulates Proliferation and Anchorage-Independent Growth by Directly Targeting Tob2 in Hepatocellular Carcinoma.
Journal of cellular biochemistry (2015-02-05)
Hongxing Shen, Wenjuan Li, Yuan Tian, Peng Xu, Hui Wang, Jinxiang Zhang, Youjun Li
要旨
The dysregulation of microRNAs (miRNAs) contributes to the pathogenesis of human malignancies, and miRNA expression can be affected by genetic and epigenetic changes, such as methylation of the CpG islands of their promoters. To identify miRNAs regulated by DNA methylation, the global miRNA expression profile was analyzed in two hepatocellular carcinoma (HCC) cell lines and two normal immortalized cell lines treated with 5-Aza-2'-deoxycytidine (DAC, an inhibitor of DNA methylation) plus TSA (Trichostatin A, histone deacetylase inhibitor). Results revealed that these epigenetic drugs differentially affect miRNA expression that is dependent or independent of cell type, especially miR-362-3p. miR-362-3p expression increased while methylation of its promoter significantly decreased in human HCC cells and tissues compared with normal cells and adjacent noncancerous tissues. Ectopic expression of miR-362-3p increased proliferation and anchorage-independent soft agar growth and its expression inhibition had opposing effects that were associated with regulation of its direct target-Tob2 in HCC cells. Inhibition of Tob2 recapitulated the effects of miR-362-3p overexpression, whereas enforced Tob2 expression reversed the promoting effects of miR-362-3p. Tob2 expression was reduced in human primary HCCs compared to adjacent noncancerous tissues. Our findings suggest that dysregulation of miR-362-3p and Tob2 may contribute to HCC malignancy.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ジメチルスルホキシド, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, for molecular biology
Sigma-Aldrich
ジメチルスルホキシド, anhydrous, ≥99.9%
Sigma-Aldrich
ジメチルスルホキシド, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
ヨウ化プロピジウム, ≥94.0% (HPLC)
Sigma-Aldrich
ヨウ化プロピジウム 溶液, solution (1.0 mg/ml in water)
Sigma-Aldrich
ヒドロキノン, ReagentPlus®, ≥99%
Sigma-Aldrich
ジメチルスルホキシド, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
トリコスタチンA, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
ヒドロキノン, ReagentPlus®, 99%
Sigma-Aldrich
ジメチルスルホキシド, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
ジメチルスルホキシド, PCR Reagent
Sigma-Aldrich
ヒドロキノン, meets USP testing specifications
Sigma-Aldrich
ジメチルスルホキシド, JIS special grade, ≥99.0%
Sigma-Aldrich
ヒドロキノン, SAJ special grade, ≥99.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
ジメチルスルホキシド, suitable for HPLC
Sigma-Aldrich
ジメチルスルホキシド, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
ヒドロキノン, SAJ first grade, ≥99.0%
Sigma-Aldrich
ジメチルスルホキシド, SAJ first grade, ≥99.0%
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5%
Sigma-Aldrich
α,α-Dicyanoethyl acetate, 98%