• ホーム
  • 検索結果
  • Artesunate-enhanced apoptosis of human high-risk myelodysplastic cells induced by the DNA methyltransferase inhibitor decitabine.
Artesunate-enhanced apoptosis of human high-risk myelodysplastic cells induced by the DNA methyltransferase inhibitor decitabine.
Oncology letters (2015-07-03)
Ying Wang, Fuxu Wang, Shupeng Wen, Yujie Guo, Xuan Liu, Xuejun Zhang, Ling Pan
要旨
The present study aimed to investigate whether artesunate (ART) could enhance the rate of apoptosis induced by decitabine (DAC) in the high-risk myelodysplastic syndrome (MDS) SKM-1 cell line, and examine the potential underlying mechanisms. The cytotoxicity and effect upon the apoptosis of ART and DAC in the SKM-1 cells was detected using the cell counting kit-8 assay and flow cytometry, respectively. The SKM-1 protein expression levels of activated caspase-3, -9 and -8, cleaved poly(ADP-ribose) polymerase and apoptosis-inducing factor (AIF) were measured by western blotting. The laser confocal microscope analysis revealed AIF transfer to the nucleus. The growth inhibition and apoptosis rates of the ART- and DAC-treated SKM-1 cells were significantly increased compared with those of the single agent-treated SKM-1 cells (P<0.05). In addition, ART and DAC induced caspase-dependent apoptosis, while ART, but not DAC, induced caspase-independent apoptosis via AIF transfer from the mitochondria to the nucleus. In addition, ART-DAC-induced cell death was not attenuated by the caspase-3/7 inhibitor, Ac-DEVD-CHO. The results of the present study suggested that the ART-DAC combination exhibited increased effectiveness compared with the single-agent therapy,
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ジメチルスルホキシド, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
ジメチルスルホキシド, for molecular biology
Sigma-Aldrich
ジメチルスルホキシド, anhydrous, ≥99.9%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
ジメチルスルホキシド, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
ジメチルスルホキシド, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
ヨウ化プロピジウム, ≥94.0% (HPLC)
Sigma-Aldrich
エチレンジアミン四酢酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
フェニルメチルスルホニルフルオリド, ≥98.5% (GC)
Sigma-Aldrich
エチレンジアミン四酢酸, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, for molecular biology, ≥97.0%
Sigma-Aldrich
HEPES緩衝液, 1 M in H2O
Sigma-Aldrich
ヨウ化プロピジウム 溶液, solution (1.0 mg/ml in water)
Sigma-Aldrich
オルトバナジン酸ナトリウム, ≥90% (titration)
Sigma-Aldrich
ジメチルスルホキシド, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, ≥97.0%
Sigma-Aldrich
エチレンジアミン四酢酸, anhydrous, crystalline, BioReagent, suitable for cell culture
SAFC
HEPES
Sigma-Aldrich
エチレンジアミン四酢酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
エチレンジアミン四酢酸, purified grade, ≥98.5%, powder
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
ジメチルスルホキシド, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
フッ化ナトリウム, BioXtra, ≥99%
Sigma-Aldrich
フェニルメチルスルホニルフルオリド, ≥99.0% (T)
Sigma-Aldrich
エチレンジアミン四酢酸, 99.995% trace metals basis
Sigma-Aldrich
オルトバナジン酸ナトリウム, 99.98% trace metals basis
Sigma-Aldrich
ジメチルスルホキシド, PCR Reagent
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SAFC
HEPES