  • Search Results
  • ADAM12-directed ectodomain shedding of E-cadherin potentiates trophoblast fusion.
ADAM12-directed ectodomain shedding of E-cadherin potentiates trophoblast fusion.
Cell death and differentiation (2015-04-25)
M Aghababaei, K Hogg, S Perdu, W P Robinson, A G Beristain
ABSTRACT
Trophoblasts, placental cells of epithelial lineage, undergo extensive differentiation to form the cellular components of the placenta. Trophoblast progenitor cell differentiation into the multinucleated syncytiotrophoblast is a key developmental process required for placental function, where defects in syncytiotrophoblast formation and turnover associate with placental pathologies and link to poor pregnancy outcomes. The cellular and molecular processes governing syncytiotrophoblast formation are poorly understood, but require the activation of pathways that direct cell fusion. The protease, A Disintegrin and Metalloproteinase 12 (ADAM12), controls cell fusion in myoblasts and is highly expressed in the placenta localizing to multiple trophoblast populations. However, the importance of ADAM12 in regulating trophoblast fusion is unknown. Here, we describe a function for ADAM12 in regulating trophoblast fusion. Using two distinct trophoblast models of cell fusion, we show that ADAM12 is dynamically upregulated and is under the transcriptional control of protein kinase A. siRNA-directed loss of ADAM12 impedes spontaneous fusion of primary cytotrophoblasts, whereas overexpression of the secreted variant, ADAM12S, potentiates cell fusion in the Bewo trophoblast cell line. Mechanistically, both ectopic and endogenous levels of ADAM12 were shown to control trophoblast fusion through E-cadherin ectodomain shedding and remodeling of intercellular boundaries. This study describes a novel role for ADAM12 in placental development, specifically highlighting its importance in controlling the differentiation of villous cytotrophoblasts into multinucleated cellular structures. Moreover, this work identifies E-cadherin as a novel ADAM12 substrate, and highlights the significance that cell adhesion molecule ectodomain shedding has in normal development.
Sigma-Aldrich
グリセロール, for molecular biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
デオキシリボヌクレアーゼ I ウシ膵臓由来, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
グリセロール, ≥99.5%
Sigma-Aldrich
デオキシコール酸ナトリウム, ≥97% (titration)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ACS reagent, ≥99.0%
Sigma-Aldrich
3-イソブチル-1-メチルキサンチン, ≥99% (HPLC), powder
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
グリセロール, ACS reagent, ≥99.5%
Sigma-Aldrich
グリセロール, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
エチレンジアミン四酢酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
フェニルメチルスルホニルフルオリド, ≥98.5% (GC)
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
フォルスコリン, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
リン酸ナトリウム, 96%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
グリセロール, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
フッ化ナトリウム, ACS reagent, ≥99%
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
エチレンジアミン四酢酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
デオキシコール酸ナトリウム, BioXtra, ≥98.0% (dry matter, NT)