  • Search Results
  • Uncoupling Neogenin association with lipid rafts promotes neuronal survival and functional recovery after stroke.
Uncoupling Neogenin association with lipid rafts promotes neuronal survival and functional recovery after stroke.
Cell death & disease (2015-05-08)
A P Shabanzadeh, N G Tassew, K Szydlowska, M Tymianski, P Banerjee, R J Vigouroux, J H Eubanks, L Huang, M Geraerts, P D Koeberle, B K Mueller, P P Monnier
ABSTRACT
The dependence receptor Neogenin and its ligand, the repulsive guidance molecule a (RGMa), regulate apoptosis and axonal growth in the developing and the adult central nervous system (CNS). Here, we show that this pathway has also a critical role in neuronal death following stroke, and that providing RGMa to neurons blocks Neogenin-induced death. Interestingly, the Neogenin pro-death function following ischemic insult depends on Neogenin association with lipid rafts. Thus, a peptide that prevents Neogenin association with lipid rafts increased neuronal survival in several in vitro stroke models. In rats, a pro-survival effect was also observed in a model of ocular ischemia, as well as after middle cerebral artery occlusion (MCAO). Treatments that prevented Neogenin association with lipid rafts improved neuronal survival and the complexity of the neuronal network following occlusion of the middle artery. Toward the development of a treatment for stroke, we developed a human anti-RGMa antibody that also prevents Neogenin association with lipid rafts. We show that this antibody also protected CNS tissue from ischemic damage and that its application resulted in a significant functional improvement even when administrated 6 h after artery occlusion. Thus, our results draw attention to the role of Neogenin and lipid rafts as potential targets following stroke.
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
重炭酸ナトリウム, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
ドデシル硫酸ナトリウム, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ACS reagent, ≥99.0%
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
ドデシル硫酸ナトリウム, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
重炭酸ナトリウム, BioXtra, 99.5-100.5%
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
ドデシル硫酸ナトリウム, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥90%
Sigma-Aldrich
塩化ナトリウム, 99.999% trace metals basis
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥98.0% (GC)
Sigma-Aldrich
塩化ナトリウム, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
塩化ナトリウム, SAJ first grade, ≥99.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
重炭酸ナトリウム, Hybri-Max, powder, suitable for hybridoma, ≥99.5%