  • Search Results
  • Targeting neddylation induces DNA damage and checkpoint activation and sensitizes chronic lymphocytic leukemia B cells to alkylating agents.
Targeting neddylation induces DNA damage and checkpoint activation and sensitizes chronic lymphocytic leukemia B cells to alkylating agents.
Cell death & disease (2015-07-15)
C Paiva, J C Godbersen, A Berger, J R Brown, A V Danilov
ABSTRACT
Microenvironment-mediated upregulation of the B-cell receptor (BCR) and nuclear factor-κB (NF-κB) signaling in CLL cells resident in the lymph node and bone marrow promotes apoptosis evasion and clonal expansion. We recently reported that MLN4924 (pevonedistat), an investigational agent that inhibits the NEDD8-activating enzyme (NAE), abrogates stromal-mediated NF-κB pathway activity and CLL cell survival. However, the NAE pathway also assists degradation of multiple other substrates. MLN4924 has been shown to induce DNA damage and cell cycle arrest, but the importance of this mechanism in primary neoplastic B cells has not been studied. Here we mimicked the lymph node microenvironment using CD40 ligand (CD40L)-expressing stroma and interleukin-21 (IL-21) to find that inducing proliferation of the primary CLL cells conferred enhanced sensitivity to NAE inhibition. Treatment of the CD40-stimulated CLL cells with MLN4924 resulted in deregulation of Cdt1, a DNA replication licensing factor, and cell cycle inhibitors p21 and p27. This led to DNA damage, checkpoint activation and G2 arrest. Alkylating agents bendamustine and chlorambucil enhanced MLN4924-mediated DNA damage and apoptosis. These events were more prominent in cells stimulated with IL-21 compared with CD40L alone, indicating that, following NAE inhibition, the culture conditions were able to direct CLL cell fate from an NF-κB inhibition to a Cdt1 induction program. Our data provide insight into the biological consequences of targeting NAE in CLL and serves as further rationale for studying the clinical activity of MLN4924 in CLL, particularly in combination with alkylating agents.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
ホルムアルデヒド 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
ホルムアルデヒド 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
エチレンジアミン四酢酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
ピルビン酸ナトリウム, ReagentPlus®, ≥99%
Sigma-Aldrich
ヨウ化プロピジウム, ≥94.0% (HPLC)
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
フェニルメチルスルホニルフルオリド, ≥98.5% (GC)
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-グルタミン, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, for molecular biology, ≥97.0%
Sigma-Aldrich
リン酸ナトリウム, 96%
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
エタノール, JIS special grade, ≥99.5%
Sigma-Aldrich
エチレンジアミン四酢酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
ピルビン酸ナトリウム, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
エチルアルコール(純粋), 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
HEPES緩衝液, 1 M in H2O
Sigma-Aldrich
ヨウ化プロピジウム 溶液, solution (1.0 mg/ml in water)
Sigma-Aldrich
エチレングリコール-ビス(2-アミノエチルエーテル)-N,N,N′,N′-四酢酸, ≥97.0%
Sigma-Aldrich
L-グルタミン, ReagentPlus®, ≥99% (HPLC)
SAFC
L-グルタミン
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
エタノール, SAJ first grade, ≥99.5%
Sigma-Aldrich
エチレンジアミン四酢酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture