  • Search Results
  • Honey reduces blood alcohol concentration but not affects the level of serum MDA and GSH-Px activity in intoxicated male mice models.
Honey reduces blood alcohol concentration but not affects the level of serum MDA and GSH-Px activity in intoxicated male mice models.
BMC complementary and alternative medicine (2015-07-15)
Peiying Shi, Bing Chen, Conghai Chen, Jingyang Xu, Zhenhuang Shen, Xiaoqing Miao, Hong Yao
ABSTRACT
For a long time, honey was purportedly helpful to prevent drunkenness and relieve hangover symptoms. However, few of the assertions have experienced scientific assessment. The present study examined the effects of honey on intoxicated male mice. Low or high doses of lychee flower honey (2.19 or 4.39 g/kg body weight, respectively) were single orally administrated 30 min before the ethanol intoxication of mice, followed by recording the locomotor activity by autonomic activity instrument and observing the climbing ability after alcohol. On the other hand, 2.19 g/kg honey was single orally administrated 5 min after the ethanol intoxication of mice, followed by determining the ethanol concentration in mice blood. In addition, subacute alcoholism mice models were developed and after the treatment of 2.19 g/kg honey s.i.d for successive three days, the level of serum malondialdehyde (MDA) and glutathione peroxidase (GSH-Px) activity were detected in the models. Both of the two doses of honey increased the autonomic activity of alcoholized mice. Furthermore, the treatment of 2.19 g/kg honey could decrease significantly the blood ethanol concentration in intoxicated mice. The anti-intoxication activity of honey could be due to the effect of the fructose contained in the honey. Meanwhile, honey could not affect the serum MDA level and GSH-Px activity in alcoholism mice models. Honey indeed possesses anti-intoxication activity.
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
4,4′-ジアミノジフェニルメタン, ≥97.0% (GC)
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
塩化ナトリウム, 99.999% trace metals basis
Sigma-Aldrich
塩化ナトリウム, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
塩化ナトリウム, SAJ first grade, ≥99.0%
Sigma-Aldrich
塩化ナトリウム, tablet
Sigma-Aldrich
塩化ナトリウム, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
塩化ナトリウム 溶液, 1 M
Sigma-Aldrich
塩化ナトリウム, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
塩化ナトリウム, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
塩化ナトリウム 溶液, 0.1 M
Sigma-Aldrich
塩化ナトリウム-35Cl, 99 atom % 35Cl