  • Search Results
  • drd2-cre:ribotag mouse line unravels the possible diversity of dopamine d2 receptor-expressing cells of the dorsal mouse hippocampus.
drd2-cre:ribotag mouse line unravels the possible diversity of dopamine d2 receptor-expressing cells of the dorsal mouse hippocampus.
Hippocampus (2014-12-30)
Emma Puighermanal, Anne Biever, Julie Espallergues, Giuseppe Gangarossa, Dimitri De Bundel, Emmanuel Valjent
ABSTRACT
Increasing evidences suggest that dopamine facilitates the encoding of novel memories by the hippocampus. However, the role of dopamine D2 receptors (D2R) in such regulations remains elusive due to the lack of the precise identification of hippocampal D2R-expressing cells. To address this issue, mice expressing the ribosomal protein Rpl22 tagged with the hemagglutinin (HA) epitope were crossed with Drd2-Cre mice allowing the selective expression of HA in D2R-containing cells (Drd2-Cre:RiboTag mice). This new transgenic model revealed a more widespread pattern of D2R-expressing cells identified by HA immunoreactivity than the one initially reported in Drd2-EGFP mice, in which the hilar mossy cells were the main neuronal population detectable. In Drd2-Cre:RiboTag mice, scattered HA/GAD67-positive neurons were detected throughout the CA1/CA3 subfields, being preferentially localized in stratum oriens and stratum lacunosum-moleculare. At the cellular level, HA-labeled cells located in CA1/CA3 subfields co-localized with calcium-binding proteins (parvalbumin, calbindin, and calretinin), neuropeptides (neuropeptide Y, somatostatin), and other markers (neuronal nitric oxide synthase, mGluR1α, reelin, coupTFII, and potassium channel-interacting protein 1). These results suggest that in addition to the glutamatergic hilar mossy cells, D2R-expressing cells constitute a subpopulation of GABAergic hippocampal interneurons.
Sigma-Aldrich
エチレングリコール, anhydrous, 99.8%
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
1,4-ジアザビシクロ[2.2.2]オクタン, ReagentPlus®, ≥99%
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
オクタン, anhydrous, ≥99%
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%
Sigma-Aldrich
DABCO® 33-LV
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
塩化ナトリウム, 99.999% trace metals basis
Sigma-Aldrich
エチレングリコール, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
キシラジン, ≥99%
Sigma-Aldrich
塩化ナトリウム, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
塩化ナトリウム, SAJ first grade, ≥99.0%
Sigma-Aldrich
塩化ナトリウム, tablet
Sigma-Aldrich
エチレングリコール 溶液
Sigma-Aldrich
塩化ナトリウム, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
エチレングリコール 溶液, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
トリス(tert-ブトキシ)シラノール, 99.999%
Sigma-Aldrich
塩化ナトリウム 溶液, 1 M
Sigma-Aldrich
塩化ナトリウム, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
塩化ナトリウム, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
オクタン, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
エチレングリコール, JIS special grade, ≥99.5%