  • Search Results
  • Cloning, expression, purification, crystallization and preliminary X-ray diffraction studies of NAD synthetase from methicillin-resistant Staphylococcus aureus.
Cloning, expression, purification, crystallization and preliminary X-ray diffraction studies of NAD synthetase from methicillin-resistant Staphylococcus aureus.
Acta crystallographica. Section F, Structural biology communications (2015-06-10)
Gajanan Kashinathrao Arbade, Sandeep Kumar Srivastava
ABSTRACT
Staphylococcus aureus is an important human and animal pathogen that causes a wide range of infections. The prevalence of multidrug-resistant S. aureus strains in both hospital and community settings makes it imperative to characterize new drug targets to combat S. aureus infections. In this context, enzymes involved in NAD metabolism and synthesis are significant drug targets as NAD is a central player in several cellular processes. NAD synthetase catalyzes the last step in the biosynthesis of nicotinamide adenine dinucleotide, making it a crucial intermediate enzyme linked to the biosynthesis of several amino acids, purine and pyrimidine nucleotides, coenzymes and antibiotics.
Sigma-Aldrich
グリセロール, for molecular biology, ≥99.0%
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
グリセロール, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
グリセロール, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
グリセリン, meets USP testing specifications
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
グリセロール 溶液, 83.5-89.5% (T)
Sigma-Aldrich
グリセロール, BioXtra, ≥99% (GC)
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
グリセロール, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
グリセロール, FCC, FG
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
塩化ナトリウム, 99.999% trace metals basis
Sigma-Aldrich
塩化ナトリウム, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
塩化ナトリウム, SAJ first grade, ≥99.0%
Sigma-Aldrich
塩化ナトリウム, tablet
Sigma-Aldrich
グリセロール, SAJ first grade, ≥98.0%
Sigma-Aldrich
グリセロール, JIS special grade, ≥99.0%
Sigma-Aldrich
塩化ナトリウム, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
塩化ナトリウム 溶液, 1 M
Sigma-Aldrich
塩化ナトリウム, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
塩化ナトリウム, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
塩化ナトリウム 溶液, 0.1 M