  • Search Results
  • Enhancement of SOX-2 expression and ROS accumulation by culture of A172 glioblastoma cells under non-adherent culture conditions.
Enhancement of SOX-2 expression and ROS accumulation by culture of A172 glioblastoma cells under non-adherent culture conditions.
Oncology reports (2015-06-03)
Chang-Nim Im, Hye Hyeon Yun, Hyung Jae Yoo, Myung-Jin Park, Jeong-Hwa Lee
ABSTRACT
More efficient isolation and identification of cancer stem cells (CSCs) would help in determining their fundamental roles in tumor biology. The classical tool for this purpose is anchorage-independent tumorsphere culture. We compared the effects of differently textured culture plates and serum deprivation on the acquisition of CSC properties of A172 glioblastoma cells. Cells were cultured on standard polystyrene-treated plates, ultra-low attachment, poly (2-hydroxyethyl methacrylate)-coated plates, and 1% agar-coated plates with 10% serum or in serum-free glioblastoma sphere medium (GBM). Based on mitochondrial reductase activity and subG1 proportions, non-adherent conditions had a greater impact on A172 cell viability than serum deprivation. Among the stemness-related genes, SOX-2 expression was significantly upregulated by serum deprivation under non-adherent conditions, while several epithelial-to-mesenchymal transition (EMT)-related genes were less dependent on serum. In addition, reactive oxygen species (ROS) accumulation in A172 cells was significantly increased in GBM under non-adherent conditions. Despite the correlation between SOX-2 induction and ROS accumulation, treatment with the ROS scavenger N-acetyl-l-cysteine did not prevent SOX-2 expression, suggesting that ROS accumulation is not an essential requirement for induction of SOX-2. Our results suggested that cultivation of cancer cells under conditions of serum deprivation in an anchorage-independent manner may enrich SOX-2-expressing CSC-like cells in vitro.
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
デオキシコール酸ナトリウム, ≥97% (titration)
Sigma-Aldrich
N-アセチル-L-システイン, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
ドデシル硫酸ナトリウム, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ACS reagent, ≥99.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
N-アセチル-L-システイン, BioReagent, suitable for cell culture
Sigma-Aldrich
ヨウ化プロピジウム, ≥94.0% (HPLC)
Sigma-Aldrich
2',7'-ジクロロフルオレセイン二酢酸, ≥97%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
デオキシコール酸ナトリウム, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
ヨウ化プロピジウム 溶液, solution (1.0 mg/ml in water)
Sigma-Aldrich
ドデシル硫酸ナトリウム, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム 溶液, BioUltra, for molecular biology, 20% in H2O
SAFC
デオキシコール酸ナトリウム
Supelco
ドデシル硫酸ナトリウム, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
N-アセチル-L-システイン, BioXtra, ≥99% (TLC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥90%
Sigma-Aldrich
ビシンコニン酸 二ナトリウム塩 水和物, ≥98% (HPLC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥98.0% (GC)
Sigma-Aldrich
ドデシル硫酸ナトリウム, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
ドデシル硫酸ナトリウム, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
ドデシル硫酸ナトリウム, SAJ special grade, ≥97.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, SAJ first grade, ≥95.0%
Sigma-Aldrich
ドデシル硫酸ナトリウム, ≥99.0%
Sigma-Aldrich
N-アセチル-L-システイン, SAJ special grade, 98.0-102.0%
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)